Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10799991-10800232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4G2
Entrez Gene ID
1982
Gene Name
eukaryotic translation initiation factor 4 gamma 2
Gene Aliases
AAG1, DAP5, NAT1, P97
UniGene
Hs.183684
Gene Chromosome Location
Chr.11: 10797046-10809035 on Build GRCh38
Transcripts
NM_001042559.2 NM_001172705.1 NM_001418.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
415351 c.1995A>G p.L665L Chr.11: 10800214-10800214 on Build GRCh38
6859765 c.2035C>T p.Q679* Chr.11: 10800174-10800174 on Build GRCh38
6067428 c.2115C>G p.L705L Chr.11: 10800094-10800094 on Build GRCh38
5892125 c.1985C>A p.A662D Chr.11: 10800224-10800224 on Build GRCh38
415352 c.2014C>A p.P672T Chr.11: 10800195-10800195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201624742 Chr.11: 10800063-10800063 on Build GRCh38
rs759140547 Chr.11: 10800157-10800157 on Build GRCh38
rs771175972 Chr.11: 10800062-10800062 on Build GRCh38
rs766059933 Chr.11: 10800141-10800141 on Build GRCh38
rs201735345 Chr.11: 10800055-10800055 on Build GRCh38
rs73417799 Chr.11: 10800068-10800068 on Build GRCh38
rs768966280 Chr.11: 10800097-10800097 on Build GRCh38
rs748216750 Chr.11: 10800054-10800055 on Build GRCh38
rs753683796 Chr.11: 10800202-10800202 on Build GRCh38
rs752766878 Chr.11: 10800047-10800047 on Build GRCh38
rs146119644 Chr.11: 10800150-10800150 on Build GRCh38
rs778947703 Chr.11: 10800064-10800064 on Build GRCh38
rs938085863 Chr.11: 10800034-10800034 on Build GRCh38
rs888546932 Chr.11: 10800067-10800067 on Build GRCh38
rs201263738 Chr.11: 10800203-10800203 on Build GRCh38
rs749541651 Chr.11: 10800061-10800061 on Build GRCh38
rs762024442 Chr.11: 10800132-10800132 on Build GRCh38
rs776023527 Chr.11: 10800074-10800074 on Build GRCh38
rs774106928 Chr.11: 10800149-10800149 on Build GRCh38
rs1051389824 Chr.11: 10800059-10800059 on Build GRCh38
rs181932480 Chr.11: 10800044-10800044 on Build GRCh38
rs143019826 Chr.11: 10800050-10800050 on Build GRCh38
rs758608139 Chr.11: 10800063-10800063 on Build GRCh38
rs752723987 Chr.11: 10800180-10800180 on Build GRCh38
rs747481512 Chr.11: 10800077-10800077 on Build GRCh38
rs752634255 Chr.11: 10800058-10800058 on Build GRCh38
rs763856120 Chr.11: 10800200-10800200 on Build GRCh38
rs767114888 Chr.11: 10800175-10800175 on Build GRCh38
rs777039041 Chr.11: 10800129-10800129 on Build GRCh38
rs777595979 Chr.11: 10800060-10800060 on Build GRCh38
rs772062949 Chr.11: 10800058-10800062 on Build GRCh38
rs551650385 Chr.11: 10800049-10800049 on Build GRCh38
rs756119421 Chr.11: 10800186-10800186 on Build GRCh38
rs750079787 Chr.11: 10800042-10800042 on Build GRCh38
rs778824877 Chr.11: 10800052-10800053 on Build GRCh38
rs766669437 Chr.11: 10800073-10800073 on Build GRCh38
rs905233016 Chr.11: 10800032-10800032 on Build GRCh38
rs754471437 Chr.11: 10800054-10800054 on Build GRCh38
rs772086746 Chr.11: 10800072-10800072 on Build GRCh38
rs764686764 Chr.11: 10800064-10800067 on Build GRCh38
rs768969714 Chr.11: 10800047-10800049 on Build GRCh38

View Full Product Details