Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41894252-41894495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF1
Entrez Gene ID
9138
Gene Name
Rho guanine nucleotide exchange factor 1
Gene Aliases
GEF1, LBCL2, LSC, P115-RHOGEF, SUB1.5
UniGene
Hs.631550
Gene Chromosome Location
Chr.19: 41883200-41907454 on Build GRCh38
Transcripts
NM_004706.3 NM_198977.1 NM_199002.1 XM_005259386.4 XM_005259390.4 XM_011527468.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5803797 c.806G>T p.R269L Chr.19: 41894467-41894467 on Build GRCh38
7226188 c.785A>G p.K262R Chr.19: 41894302-41894302 on Build GRCh38
3404282 c.810G>A p.S270S Chr.19: 41894471-41894471 on Build GRCh38
73716 c.740G>A p.R247Q Chr.19: 41894257-41894257 on Build GRCh38
4420818 c.765G>A p.S255S Chr.19: 41894282-41894282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782722507 Chr.19: 41894404-41894405 on Build GRCh38
rs944286916 Chr.19: 41894437-41894437 on Build GRCh38
rs371929877 Chr.19: 41894406-41894406 on Build GRCh38
rs782446869 Chr.19: 41894325-41894325 on Build GRCh38
rs991215252 Chr.19: 41894382-41894382 on Build GRCh38
rs782295165 Chr.19: 41894332-41894332 on Build GRCh38
rs782629121 Chr.19: 41894475-41894475 on Build GRCh38
rs12104228 Chr.19: 41894377-41894377 on Build GRCh38
rs782517739 Chr.19: 41894313-41894313 on Build GRCh38
rs370981860 Chr.19: 41894470-41894470 on Build GRCh38
rs142729688 Chr.19: 41894463-41894463 on Build GRCh38
rs149993474 Chr.19: 41894359-41894359 on Build GRCh38
rs782043796 Chr.19: 41894412-41894412 on Build GRCh38
rs781884761 Chr.19: 41894467-41894467 on Build GRCh38
rs782059204 Chr.19: 41894336-41894336 on Build GRCh38
rs782408918 Chr.19: 41894411-41894411 on Build GRCh38
rs374677524 Chr.19: 41894354-41894354 on Build GRCh38
rs782799693 Chr.19: 41894438-41894438 on Build GRCh38
rs1014802424 Chr.19: 41894346-41894346 on Build GRCh38
rs782808751 Chr.19: 41894290-41894290 on Build GRCh38
rs782358325 Chr.19: 41894401-41894401 on Build GRCh38
rs368940793 Chr.19: 41894281-41894281 on Build GRCh38
rs546989400 Chr.19: 41894385-41894385 on Build GRCh38
rs552631968 Chr.19: 41894352-41894352 on Build GRCh38
rs1041220028 Chr.19: 41894462-41894462 on Build GRCh38
rs145191723 Chr.19: 41894403-41894403 on Build GRCh38
rs782660388 Chr.19: 41894326-41894326 on Build GRCh38
rs781910454 Chr.19: 41894301-41894301 on Build GRCh38
rs782763644 Chr.19: 41894471-41894471 on Build GRCh38
rs202000253 Chr.19: 41894474-41894474 on Build GRCh38
rs374846976 Chr.19: 41894448-41894448 on Build GRCh38
rs781874408 Chr.19: 41894473-41894473 on Build GRCh38
rs370278424 Chr.19: 41894339-41894339 on Build GRCh38
rs781860786 Chr.19: 41894312-41894312 on Build GRCh38
rs749020865 Chr.19: 41894464-41894464 on Build GRCh38
rs781956089 Chr.19: 41894439-41894439 on Build GRCh38
rs557324472 Chr.19: 41894417-41894417 on Build GRCh38
rs1035511549 Chr.19: 41894368-41894368 on Build GRCh38
rs782545099 Chr.19: 41894469-41894469 on Build GRCh38
rs188938411 Chr.19: 41894322-41894322 on Build GRCh38
rs782621639 Chr.19: 41894315-41894315 on Build GRCh38
rs539561941 Chr.19: 41894282-41894282 on Build GRCh38

View Full Product Details