Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 114324961-114325204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAMP1
Entrez Gene ID
283489
Gene Name
chromosome alignment maintaining phosphoprotein 1
Gene Aliases
C13orf8, CAMP, CHAMP, MRD40, ZNF828
UniGene
Hs.7542
Gene Chromosome Location
Chr.13: 114314490-114327328 on Build GRCh38
Transcripts
NM_001164144.1 NM_001164145.1 NM_032436.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6074507 c.1253A>C p.E418A Chr.13: 114325095-114325095 on Build GRCh38
4464851 c.1355C>T p.S452F Chr.13: 114325197-114325197 on Build GRCh38
1234675 c.1258C>T p.R420C Chr.13: 114325100-114325100 on Build GRCh38
3793160 c.1328C>T p.S443L Chr.13: 114325170-114325170 on Build GRCh38
695907 c.1309C>G p.P437A Chr.13: 114325151-114325151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529321452 Chr.13: 114325026-114325026 on Build GRCh38
rs782758094 Chr.13: 114325179-114325179 on Build GRCh38
rs138325089 Chr.13: 114325001-114325001 on Build GRCh38
rs374334158 Chr.13: 114325146-114325146 on Build GRCh38
rs797044962 Chr.13: 114325034-114325034 on Build GRCh38
rs547434000 Chr.13: 114325134-114325134 on Build GRCh38
rs781985494 Chr.13: 114325056-114325056 on Build GRCh38
rs371126156 Chr.13: 114325153-114325153 on Build GRCh38
rs1023708288 Chr.13: 114325057-114325057 on Build GRCh38
rs782128382 Chr.13: 114325065-114325065 on Build GRCh38
rs201826600 Chr.13: 114325159-114325159 on Build GRCh38
rs782752774 Chr.13: 114325100-114325100 on Build GRCh38
rs35076210 Chr.13: 114325108-114325109 on Build GRCh38
rs149624011 Chr.13: 114325051-114325051 on Build GRCh38
rs782338415 Chr.13: 114325133-114325133 on Build GRCh38
rs782283006 Chr.13: 114325108-114325108 on Build GRCh38
rs573576960 Chr.13: 114325166-114325166 on Build GRCh38
rs201708961 Chr.13: 114325151-114325151 on Build GRCh38
rs138201298 Chr.13: 114324990-114324990 on Build GRCh38
rs781847798 Chr.13: 114325101-114325101 on Build GRCh38
rs782564405 Chr.13: 114325109-114325109 on Build GRCh38
rs782510090 Chr.13: 114325107-114325107 on Build GRCh38
rs782393001 Chr.13: 114325009-114325009 on Build GRCh38
rs543967281 Chr.13: 114324995-114324995 on Build GRCh38
rs782021774 Chr.13: 114325021-114325021 on Build GRCh38
rs782782357 Chr.13: 114325074-114325074 on Build GRCh38
rs782454954 Chr.13: 114325096-114325096 on Build GRCh38
rs781895659 Chr.13: 114325086-114325086 on Build GRCh38
rs376497995 Chr.13: 114325035-114325035 on Build GRCh38
rs782386969 Chr.13: 114325144-114325144 on Build GRCh38
rs782251855 Chr.13: 114325002-114325002 on Build GRCh38
rs372468779 Chr.13: 114325168-114325168 on Build GRCh38
rs144334556 Chr.13: 114325120-114325120 on Build GRCh38
rs370703699 Chr.13: 114325043-114325043 on Build GRCh38
rs782192484 Chr.13: 114325130-114325130 on Build GRCh38

View Full Product Details