Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147212426-147212659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD1L
Entrez Gene ID
9557
Gene Name
chromodomain helicase DNA binding protein 1 like
Gene Aliases
ALC1, CHDL
UniGene
Hs.191164
Gene Chromosome Location
Chr.1: 147172804-147295766 on Build GRCh38
Transcripts
XM_017002858.1 NM_001144829.2 NM_001144830.2 NM_001461.3 XM_005272946.4 XM_005272947.4 XM_005272948.4 XM_006711244.3 XM_006711245.3 XM_011509350.2 XM_011509351.2 XM_017000801.1 XM_017000802.1

Gene Symbol
FMO5
Entrez Gene ID
2330
Gene Name
flavin containing monooxygenase 5
Gene Aliases
-
UniGene
Hs.642706
Gene Chromosome Location
Chr.1: 147183963-147225798 on Build GRCh38
Transcripts
XM_017002858.1 NM_001144829.2 NM_001144830.2 NM_001461.3 XM_005272946.4 XM_005272947.4 XM_005272948.4 XM_006711244.3 XM_006711245.3 XM_011509350.2 XM_011509351.2 XM_017000801.1 XM_017000802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
423713 c.578T>A p.I193N Chr.1: 147212445-147212445 on Build GRCh38
2121542 c.531T>G p.Y177* Chr.1: 147212492-147212492 on Build GRCh38
4809906 c.530A>G p.Y177C Chr.1: 147212493-147212493 on Build GRCh38
4899508 c.559A>C p.R187R Chr.1: 147212464-147212464 on Build GRCh38
5502304 c.544G>A p.G182R Chr.1: 147212479-147212479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17360066 Chr.1: 147212605-147212605 on Build GRCh38
rs782604962 Chr.1: 147212593-147212593 on Build GRCh38
rs782690948 Chr.1: 147212499-147212499 on Build GRCh38
rs782228735 Chr.1: 147212479-147212479 on Build GRCh38
rs782768099 Chr.1: 147212532-147212532 on Build GRCh38
rs782309093 Chr.1: 147212606-147212606 on Build GRCh38
rs782061860 Chr.1: 147212510-147212510 on Build GRCh38
rs782366526 Chr.1: 147212549-147212549 on Build GRCh38
rs782524155 Chr.1: 147212451-147212451 on Build GRCh38
rs782816480 Chr.1: 147212462-147212462 on Build GRCh38
rs782026224 Chr.1: 147212525-147212525 on Build GRCh38
rs587682197 Chr.1: 147212500-147212500 on Build GRCh38
rs781797303 Chr.1: 147212450-147212450 on Build GRCh38
rs781850993 Chr.1: 147212463-147212463 on Build GRCh38
rs781818663 Chr.1: 147212538-147212538 on Build GRCh38
rs782787091 Chr.1: 147212519-147212519 on Build GRCh38
rs200936105 Chr.1: 147212492-147212492 on Build GRCh38
rs782437664 Chr.1: 147212480-147212480 on Build GRCh38
rs782504617 Chr.1: 147212547-147212547 on Build GRCh38
rs782516404 Chr.1: 147212466-147212466 on Build GRCh38
rs782244207 Chr.1: 147212585-147212585 on Build GRCh38
rs781979585 Chr.1: 147212507-147212507 on Build GRCh38
rs1021501740 Chr.1: 147212604-147212604 on Build GRCh38
rs200650061 Chr.1: 147212604-147212604 on Build GRCh38
rs782694403 Chr.1: 147212579-147212579 on Build GRCh38
rs200888056 Chr.1: 147212570-147212570 on Build GRCh38
rs782278097 Chr.1: 147212491-147212491 on Build GRCh38
rs143647812 Chr.1: 147212506-147212506 on Build GRCh38
rs58351438 Chr.1: 147212527-147212527 on Build GRCh38
rs2297753 Chr.1: 147212563-147212563 on Build GRCh38
rs782603450 Chr.1: 147212474-147212474 on Build GRCh38
rs782487032 Chr.1: 147212541-147212541 on Build GRCh38
rs758203330 Chr.1: 147212604-147212605 on Build GRCh38
rs370710329 Chr.1: 147212449-147212449 on Build GRCh38
rs782576772 Chr.1: 147212482-147212482 on Build GRCh38
rs782699556 Chr.1: 147212544-147212544 on Build GRCh38

View Full Product Details