Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53501522-53501795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1D
Entrez Gene ID
776
Gene Name
calcium voltage-gated channel subunit alpha1 D
Gene Aliases
CACH3, CACN4, CACNL1A2, CCHL1A2, Cav1.3, PASNA, SANDD
UniGene
Hs.476358
Gene Chromosome Location
Chr.3: 53495049-53813151 on Build GRCh38
Transcripts
NM_000720.3 NM_001128839.2 NM_001128840.2 XM_005265448.3 XM_011534094.2 XM_011534096.2 XM_011534097.2 XM_011534100.2 XM_017007137.1 XM_017007138.1 XM_017007139.1 XM_017007140.1 XM_017007141.1 XM_017007142.1 XM_017007143.1 XM_017007144.1 XM_017007145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4584559 c.400T>C p.L134L Chr.3: 53501637-53501637 on Build GRCh38
5498526 c.378-3C>T p.? Chr.3: 53501612-53501612 on Build GRCh38
1047281 c.454G>T p.D152Y Chr.3: 53501691-53501691 on Build GRCh38
4584560 c.400T>C p.L134L Chr.3: 53501637-53501637 on Build GRCh38
5498525 c.378-3C>T p.? Chr.3: 53501612-53501612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs876657433 Chr.3: 53501687-53501687 on Build GRCh38
rs778624473 Chr.3: 53501766-53501766 on Build GRCh38
rs903117950 Chr.3: 53501548-53501548 on Build GRCh38
rs376856950 Chr.3: 53501549-53501549 on Build GRCh38
rs748538909 Chr.3: 53501574-53501574 on Build GRCh38
rs747506509 Chr.3: 53501764-53501764 on Build GRCh38
rs755197343 Chr.3: 53501703-53501703 on Build GRCh38
rs766191935 Chr.3: 53501690-53501690 on Build GRCh38
rs771377070 Chr.3: 53501767-53501767 on Build GRCh38
rs561965317 Chr.3: 53501554-53501554 on Build GRCh38
rs778855715 Chr.3: 53501736-53501736 on Build GRCh38
rs778387696 Chr.3: 53501763-53501763 on Build GRCh38
rs373329119 Chr.3: 53501670-53501670 on Build GRCh38
rs764813486 Chr.3: 53501757-53501757 on Build GRCh38
rs6805302 Chr.3: 53501696-53501696 on Build GRCh38
rs773439379 Chr.3: 53501585-53501585 on Build GRCh38
rs558073440 Chr.3: 53501591-53501591 on Build GRCh38
rs777726885 Chr.3: 53501557-53501557 on Build GRCh38
rs965658148 Chr.3: 53501601-53501602 on Build GRCh38
rs778977779 Chr.3: 53501567-53501567 on Build GRCh38
rs773914210 Chr.3: 53501618-53501618 on Build GRCh38
rs761648251 Chr.3: 53501619-53501619 on Build GRCh38
rs1017382807 Chr.3: 53501699-53501699 on Build GRCh38
rs111367499 Chr.3: 53501582-53501582 on Build GRCh38
rs529772123 Chr.3: 53501592-53501592 on Build GRCh38
rs769545031 Chr.3: 53501586-53501586 on Build GRCh38
rs775247704 Chr.3: 53501589-53501589 on Build GRCh38
rs760502506 Chr.3: 53501676-53501676 on Build GRCh38
rs77988100 Chr.3: 53501649-53501649 on Build GRCh38
rs138356667 Chr.3: 53501737-53501737 on Build GRCh38
rs781697668 Chr.3: 53501771-53501771 on Build GRCh38
rs749062022 Chr.3: 53501660-53501660 on Build GRCh38
rs767546714 Chr.3: 53501640-53501640 on Build GRCh38
rs1032814425 Chr.3: 53501593-53501593 on Build GRCh38

View Full Product Details