Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66940095-66940366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES2
Entrez Gene ID
8824
Gene Name
carboxylesterase 2
Gene Aliases
CE-2, CES2A1, PCE-2, iCE
UniGene
Hs.282975
Gene Chromosome Location
Chr.16: 66934444-66945096 on Build GRCh38
Transcripts
NM_003869.5 NM_198061.2 NR_036684.1 XM_011523421.2 XM_017023818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6283013 c.654C>T p.G218G Chr.16: 66940260-66940260 on Build GRCh38
1736306 c.621G>A p.M207I Chr.16: 66940227-66940227 on Build GRCh38
6273124 c.733G>T p.V245F Chr.16: 66940339-66940339 on Build GRCh38
6627399 c.690C>A p.A230A Chr.16: 66940296-66940296 on Build GRCh38
4061714 c.644T>C p.L215P Chr.16: 66940250-66940250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373489723 Chr.16: 66940189-66940189 on Build GRCh38
rs756704519 Chr.16: 66940171-66940171 on Build GRCh38
rs761959207 Chr.16: 66940306-66940306 on Build GRCh38
rs777666645 Chr.16: 66940325-66940325 on Build GRCh38
rs760010628 Chr.16: 66940232-66940232 on Build GRCh38
rs756754102 Chr.16: 66940329-66940329 on Build GRCh38
rs565364625 Chr.16: 66940331-66940331 on Build GRCh38
rs368103444 Chr.16: 66940305-66940305 on Build GRCh38
rs200227138 Chr.16: 66940178-66940178 on Build GRCh38
rs745571678 Chr.16: 66940248-66940248 on Build GRCh38
rs763302523 Chr.16: 66940182-66940182 on Build GRCh38
rs375344432 Chr.16: 66940247-66940247 on Build GRCh38
rs139864573 Chr.16: 66940240-66940240 on Build GRCh38
rs934259624 Chr.16: 66940164-66940164 on Build GRCh38
rs750658371 Chr.16: 66940200-66940200 on Build GRCh38
rs746492227 Chr.16: 66940243-66940243 on Build GRCh38
rs751557530 Chr.16: 66940328-66940328 on Build GRCh38
rs767814049 Chr.16: 66940197-66940197 on Build GRCh38
rs573993430 Chr.16: 66940188-66940188 on Build GRCh38
rs758230050 Chr.16: 66940324-66940324 on Build GRCh38
rs948763121 Chr.16: 66940249-66940249 on Build GRCh38
rs72547532 Chr.16: 66940222-66940222 on Build GRCh38
rs778508138 Chr.16: 66940228-66940228 on Build GRCh38
rs763428660 Chr.16: 66940286-66940286 on Build GRCh38
rs11568312 Chr.16: 66940291-66940291 on Build GRCh38
rs150573301 Chr.16: 66940239-66940239 on Build GRCh38
rs201228525 Chr.16: 66940257-66940257 on Build GRCh38
rs766649453 Chr.16: 66940315-66940315 on Build GRCh38
rs765576852 Chr.16: 66940175-66940175 on Build GRCh38
rs775658664 Chr.16: 66940280-66940280 on Build GRCh38
rs769440021 Chr.16: 66940323-66940323 on Build GRCh38
rs764710237 Chr.16: 66940184-66940184 on Build GRCh38
rs1045963447 Chr.16: 66940330-66940330 on Build GRCh38
rs534598744 Chr.16: 66940172-66940172 on Build GRCh38
rs760754527 Chr.16: 66940311-66940311 on Build GRCh38
rs759924440 Chr.16: 66940174-66940174 on Build GRCh38
rs565571128 Chr.16: 66940143-66940143 on Build GRCh38
rs183965659 Chr.16: 66940125-66940125 on Build GRCh38
rs202022546 Chr.16: 66940227-66940227 on Build GRCh38
rs202072951 Chr.16: 66940309-66940309 on Build GRCh38

View Full Product Details