Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121763726-121763995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAB2IP
Entrez Gene ID
153090
Gene Name
DAB2 interacting protein
Gene Aliases
AF9Q34, AIP-1, AIP1, DIP1/2
UniGene
Hs.522378
Gene Chromosome Location
Chr.9: 121566883-121785530 on Build GRCh38
Transcripts
NM_032552.3 NM_138709.2 XM_005251719.4 XM_005251721.1 XM_005251724.4 XM_011518264.2 XM_011518265.2 XM_011518266.2 XM_011518267.2 XM_011518270.2 XM_011518271.2 XM_017014298.1 XM_017014299.1 XM_017014300.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6114769 c.1300G>A p.A434T Chr.9: 121763803-121763803 on Build GRCh38
7363623 c.1379C>T p.S460L Chr.9: 121763798-121763798 on Build GRCh38
6639789 c.1357G>A p.E453K Chr.9: 121763776-121763776 on Build GRCh38
6639787 c.1273G>A p.E425K Chr.9: 121763776-121763776 on Build GRCh38
5000045 c.1007C>T p.S336L Chr.9: 121763798-121763798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758422484 Chr.9: 121763888-121763888 on Build GRCh38
rs371930308 Chr.9: 121763750-121763750 on Build GRCh38
rs751443590 Chr.9: 121763895-121763895 on Build GRCh38
rs759469821 Chr.9: 121763891-121763891 on Build GRCh38
rs926537043 Chr.9: 121763886-121763886 on Build GRCh38
rs536102830 Chr.9: 121763962-121763962 on Build GRCh38
rs376330730 Chr.9: 121763841-121763841 on Build GRCh38
rs190350865 Chr.9: 121763913-121763913 on Build GRCh38
rs769633331 Chr.9: 121763812-121763812 on Build GRCh38
rs761118729 Chr.9: 121763847-121763847 on Build GRCh38
rs899933252 Chr.9: 121763920-121763920 on Build GRCh38
rs747964485 Chr.9: 121763906-121763906 on Build GRCh38
rs985379803 Chr.9: 121763787-121763787 on Build GRCh38
rs551225800 Chr.9: 121763887-121763887 on Build GRCh38
rs758299546 Chr.9: 121763792-121763792 on Build GRCh38
rs781024010 Chr.9: 121763903-121763903 on Build GRCh38
rs757425324 Chr.9: 121763771-121763771 on Build GRCh38
rs777156640 Chr.9: 121763878-121763878 on Build GRCh38
rs77220406 Chr.9: 121763796-121763796 on Build GRCh38
rs200491251 Chr.9: 121763915-121763915 on Build GRCh38
rs199616778 Chr.9: 121763908-121763908 on Build GRCh38
rs139934832 Chr.9: 121763844-121763844 on Build GRCh38
rs373776876 Chr.9: 121763799-121763799 on Build GRCh38
rs754881793 Chr.9: 121763897-121763897 on Build GRCh38
rs996837187 Chr.9: 121763930-121763930 on Build GRCh38
rs773739774 Chr.9: 121763921-121763921 on Build GRCh38
rs375557765 Chr.9: 121763802-121763802 on Build GRCh38
rs80248526 Chr.9: 121763912-121763912 on Build GRCh38
rs769543291 Chr.9: 121763910-121763910 on Build GRCh38
rs761152891 Chr.9: 121763751-121763751 on Build GRCh38
rs368910721 Chr.9: 121763852-121763852 on Build GRCh38
rs1040725295 Chr.9: 121763947-121763947 on Build GRCh38
rs376373696 Chr.9: 121763752-121763752 on Build GRCh38
rs1009358761 Chr.9: 121763945-121763945 on Build GRCh38
rs539329547 Chr.9: 121763865-121763865 on Build GRCh38
rs775152978 Chr.9: 121763790-121763790 on Build GRCh38
rs778950978 Chr.9: 121763775-121763775 on Build GRCh38
rs900803727 Chr.9: 121763960-121763960 on Build GRCh38
rs145965964 Chr.9: 121763815-121763815 on Build GRCh38
rs754053381 Chr.9: 121763756-121763756 on Build GRCh38
rs926402107 Chr.9: 121763755-121763755 on Build GRCh38
rs141706892 Chr.9: 121763803-121763803 on Build GRCh38
rs76191175 Chr.9: 121763798-121763798 on Build GRCh38
rs762594726 Chr.9: 121763829-121763829 on Build GRCh38
rs574866702 Chr.9: 121763929-121763929 on Build GRCh38

View Full Product Details