Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100155317-100155560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf43
Entrez Gene ID
55262
Gene Name
chromosome 7 open reading frame 43
Gene Aliases
-
UniGene
Hs.533139
Gene Chromosome Location
Chr.7: 100154420-100158721 on Build GRCh38
Transcripts
NM_001303470.1 NM_018275.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1093987 c.1438G>A p.V480M Chr.7: 100155413-100155413 on Build GRCh38
1623390 c.1418A>C p.K473T Chr.7: 100155433-100155433 on Build GRCh38
7358704 c.1474C>G p.R492G Chr.7: 100155377-100155377 on Build GRCh38
1093988 c.1432G>A p.A478T Chr.7: 100155419-100155419 on Build GRCh38
6416128 c.1507G>A p.D503N Chr.7: 100155344-100155344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774869634 Chr.7: 100155505-100155514 on Build GRCh38
rs544128104 Chr.7: 100155420-100155420 on Build GRCh38
rs561257336 Chr.7: 100155342-100155342 on Build GRCh38
rs576474905 Chr.7: 100155419-100155419 on Build GRCh38
rs745841608 Chr.7: 100155381-100155381 on Build GRCh38
rs919794829 Chr.7: 100155393-100155393 on Build GRCh38
rs980701374 Chr.7: 100155414-100155414 on Build GRCh38
rs376067689 Chr.7: 100155435-100155435 on Build GRCh38
rs754643547 Chr.7: 100155413-100155413 on Build GRCh38
rs2272337 Chr.7: 100155498-100155498 on Build GRCh38
rs1006921376 Chr.7: 100155351-100155351 on Build GRCh38
rs747613585 Chr.7: 100155432-100155432 on Build GRCh38
rs373416018 Chr.7: 100155388-100155388 on Build GRCh38
rs370447712 Chr.7: 100155376-100155376 on Build GRCh38
rs953568981 Chr.7: 100155335-100155335 on Build GRCh38
rs753886568 Chr.7: 100155347-100155347 on Build GRCh38
rs761727928 Chr.7: 100155488-100155488 on Build GRCh38
rs776701829 Chr.7: 100155487-100155487 on Build GRCh38
rs533277014 Chr.7: 100155460-100155460 on Build GRCh38
rs973059328 Chr.7: 100155377-100155377 on Build GRCh38
rs917973550 Chr.7: 100155405-100155405 on Build GRCh38
rs768815663 Chr.7: 100155484-100155484 on Build GRCh38
rs777505392 Chr.7: 100155458-100155458 on Build GRCh38
rs970795574 Chr.7: 100155406-100155406 on Build GRCh38
rs757802165 Chr.7: 100155350-100155350 on Build GRCh38
rs779083600 Chr.7: 100155524-100155524 on Build GRCh38
rs763494345 Chr.7: 100155494-100155494 on Build GRCh38
rs780551163 Chr.7: 100155396-100155396 on Build GRCh38
rs370303096 Chr.7: 100155463-100155463 on Build GRCh38
rs372335034 Chr.7: 100155375-100155375 on Build GRCh38
rs760929714 Chr.7: 100155476-100155476 on Build GRCh38
rs141927287 Chr.7: 100155470-100155470 on Build GRCh38
rs945845318 Chr.7: 100155459-100155459 on Build GRCh38
rs758756246 Chr.7: 100155395-100155395 on Build GRCh38
rs972278965 Chr.7: 100155404-100155404 on Build GRCh38
rs866351198 Chr.7: 100155446-100155446 on Build GRCh38
rs770693224 Chr.7: 100155465-100155465 on Build GRCh38

View Full Product Details