Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8511718-8511961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF414
Entrez Gene ID
84330
Gene Name
zinc finger protein 414
Gene Aliases
ZFP414
UniGene
Hs.515114
Gene Chromosome Location
Chr.19: 8510578-8514164 on Build GRCh38
Transcripts
NM_001146175.1 NM_032370.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3371566 c.630C>T p.P210P Chr.19: 8511861-8511861 on Build GRCh38
5549180 c.693G>A p.A231A Chr.19: 8511798-8511798 on Build GRCh38
359243 c.735C>T p.P245P Chr.19: 8511756-8511756 on Build GRCh38
4743756 c.597G>A p.A199A Chr.19: 8511894-8511894 on Build GRCh38
6359802 c.736G>A p.A246T Chr.19: 8511755-8511755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771812176 Chr.19: 8511903-8511903 on Build GRCh38
rs536650276 Chr.19: 8511810-8511810 on Build GRCh38
rs771971979 Chr.19: 8511828-8511828 on Build GRCh38
rs766467703 Chr.19: 8511934-8511934 on Build GRCh38
rs867989279 Chr.19: 8511751-8511751 on Build GRCh38
rs763014445 Chr.19: 8511857-8511857 on Build GRCh38
rs922678831 Chr.19: 8511840-8511840 on Build GRCh38
rs753042081 Chr.19: 8511871-8511871 on Build GRCh38
rs868270747 Chr.19: 8511763-8511763 on Build GRCh38
rs763067451 Chr.19: 8511930-8511930 on Build GRCh38
rs200518876 Chr.19: 8511745-8511745 on Build GRCh38
rs778760286 Chr.19: 8511895-8511895 on Build GRCh38
rs965357659 Chr.19: 8511892-8511892 on Build GRCh38
rs918838798 Chr.19: 8511925-8511925 on Build GRCh38
rs914155209 Chr.19: 8511797-8511797 on Build GRCh38
rs770767844 Chr.19: 8511801-8511801 on Build GRCh38
rs1020821374 Chr.19: 8511817-8511817 on Build GRCh38
rs757407418 Chr.19: 8511739-8511739 on Build GRCh38
rs958486384 Chr.19: 8511811-8511811 on Build GRCh38
rs776598591 Chr.19: 8511926-8511926 on Build GRCh38
rs751985264 Chr.19: 8511782-8511782 on Build GRCh38
rs11554506 Chr.19: 8511875-8511875 on Build GRCh38
rs1032558721 Chr.19: 8511866-8511866 on Build GRCh38
rs1037606489 Chr.19: 8511887-8511887 on Build GRCh38
rs751755979 Chr.19: 8511865-8511865 on Build GRCh38
rs774145585 Chr.19: 8511805-8511805 on Build GRCh38
rs775161387 Chr.19: 8511907-8511907 on Build GRCh38
rs931511521 Chr.19: 8511931-8511931 on Build GRCh38
rs759446097 Chr.19: 8511824-8511824 on Build GRCh38
rs1050253282 Chr.19: 8511742-8511742 on Build GRCh38
rs750526535 Chr.19: 8511856-8511856 on Build GRCh38
rs571114437 Chr.19: 8511792-8511792 on Build GRCh38
rs980932171 Chr.19: 8511788-8511788 on Build GRCh38
rs759754516 Chr.19: 8511941-8511941 on Build GRCh38
rs375198071 Chr.19: 8511787-8511787 on Build GRCh38
rs1006487024 Chr.19: 8511881-8511881 on Build GRCh38
rs1041470351 Chr.19: 8511820-8511820 on Build GRCh38
rs777156437 Chr.19: 8511795-8511795 on Build GRCh38
rs985927589 Chr.19: 8511872-8511872 on Build GRCh38
rs748778876 Chr.19: 8511796-8511796 on Build GRCh38
rs756596910 Chr.19: 8511874-8511874 on Build GRCh38
rs745538052 Chr.19: 8511896-8511896 on Build GRCh38
rs1006105412 Chr.19: 8511746-8511746 on Build GRCh38
rs755140044 Chr.19: 8511867-8511867 on Build GRCh38
rs1049809484 Chr.19: 8511860-8511860 on Build GRCh38
rs765254777 Chr.19: 8511744-8511744 on Build GRCh38
rs758562459 Chr.19: 8511768-8511768 on Build GRCh38
rs751594662 Chr.19: 8511939-8511939 on Build GRCh38
rs973012991 Chr.19: 8511912-8511912 on Build GRCh38
rs755301704 Chr.19: 8511783-8511783 on Build GRCh38
rs945376459 Chr.19: 8511798-8511798 on Build GRCh38
rs775306654 Chr.19: 8511832-8511832 on Build GRCh38
rs760505125 Chr.19: 8511908-8511908 on Build GRCh38
rs550029086 Chr.19: 8511918-8511918 on Build GRCh38
rs926737711 Chr.19: 8511791-8511791 on Build GRCh38
rs748819124 Chr.19: 8511882-8511882 on Build GRCh38
rs770535139 Chr.19: 8511890-8511890 on Build GRCh38
rs766626003 Chr.19: 8511861-8511861 on Build GRCh38
rs764033116 Chr.19: 8511838-8511838 on Build GRCh38
rs7175 Chr.19: 8511786-8511786 on Build GRCh38
rs760561351 Chr.19: 8511834-8511834 on Build GRCh38
rs780239913 Chr.19: 8511779-8511779 on Build GRCh38
rs751852731 Chr.19: 8511859-8511859 on Build GRCh38
rs781437718 Chr.19: 8511868-8511868 on Build GRCh38

View Full Product Details