Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150272705-150272978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APH1A
Entrez Gene ID
51107
Gene Name
aph-1 homolog A, gamma-secretase subunit
Gene Aliases
6530402N02Rik, APH-1, APH-1A, CGI-78
UniGene
Hs.108408
Gene Chromosome Location
Chr.1: 150265399-150282291 on Build GRCh38
Transcripts
XM_017001417.1 NM_001301039.1 NM_001301040.1 NM_001301041.1 NM_001301042.1 NM_024579.3 XM_011509984.2

Gene Symbol
C1orf54
Entrez Gene ID
79630
Gene Name
chromosome 1 open reading frame 54
Gene Aliases
-
UniGene
Hs.91283
Gene Chromosome Location
Chr.1: 150272284-150280927 on Build GRCh38
Transcripts
XM_017001417.1 NM_001301039.1 NM_001301040.1 NM_001301041.1 NM_001301042.1 NM_024579.3 XM_011509984.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5887540 c.10C>T p.L4F Chr.1: 150272827-150272827 on Build GRCh38
1198454 c.2T>A p.M1K Chr.1: 150272819-150272819 on Build GRCh38
5887541 c.10C>T p.L4F Chr.1: 150272827-150272827 on Build GRCh38
7358304 c.24C>G p.I8M Chr.1: 150272841-150272841 on Build GRCh38
6454655 c.7G>A p.V3I Chr.1: 150272824-150272824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199961326 Chr.1: 150272884-150272884 on Build GRCh38
rs782226732 Chr.1: 150272827-150272827 on Build GRCh38
rs199601572 Chr.1: 150272774-150272774 on Build GRCh38
rs201581065 Chr.1: 150272823-150272823 on Build GRCh38
rs782570702 Chr.1: 150272839-150272839 on Build GRCh38
rs781887680 Chr.1: 150272874-150272874 on Build GRCh38
rs369066637 Chr.1: 150272872-150272872 on Build GRCh38
rs941271739 Chr.1: 150272753-150272753 on Build GRCh38
rs781825407 Chr.1: 150272892-150272892 on Build GRCh38
rs782308109 Chr.1: 150272833-150272833 on Build GRCh38
rs587735018 Chr.1: 150272778-150272778 on Build GRCh38
rs35306203 Chr.1: 150272864-150272865 on Build GRCh38
rs982981474 Chr.1: 150272749-150272749 on Build GRCh38
rs782485052 Chr.1: 150272902-150272902 on Build GRCh38
rs782601502 Chr.1: 150272824-150272824 on Build GRCh38
rs937647307 Chr.1: 150272850-150272850 on Build GRCh38
rs782741739 Chr.1: 150272786-150272786 on Build GRCh38
rs782140588 Chr.1: 150272861-150272861 on Build GRCh38
rs782818972 Chr.1: 150272736-150272736 on Build GRCh38
rs782409613 Chr.1: 150272854-150272854 on Build GRCh38
rs782268473 Chr.1: 150272848-150272848 on Build GRCh38
rs782351557 Chr.1: 150272862-150272862 on Build GRCh38
rs781977742 Chr.1: 150272863-150272863 on Build GRCh38
rs782055972 Chr.1: 150272868-150272868 on Build GRCh38
rs782564967 Chr.1: 150272904-150272904 on Build GRCh38
rs201064735 Chr.1: 150272885-150272885 on Build GRCh38
rs35018366 Chr.1: 150272844-150272845 on Build GRCh38
rs781992672 Chr.1: 150272859-150272859 on Build GRCh38
rs782087081 Chr.1: 150272875-150272876 on Build GRCh38
rs922648461 Chr.1: 150272739-150272739 on Build GRCh38
rs73011383 Chr.1: 150272921-150272921 on Build GRCh38
rs782674927 Chr.1: 150272793-150272793 on Build GRCh38
rs781793103 Chr.1: 150272787-150272787 on Build GRCh38
rs933857123 Chr.1: 150272748-150272748 on Build GRCh38
rs908473338 Chr.1: 150272750-150272750 on Build GRCh38
rs368023952 Chr.1: 150272788-150272788 on Build GRCh38
rs781858299 Chr.1: 150272809-150272809 on Build GRCh38
rs975509981 Chr.1: 150272737-150272737 on Build GRCh38

View Full Product Details