Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78588151-78588394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA5
Entrez Gene ID
1138
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases
LNCR2
UniGene
Hs.1614
Gene Chromosome Location
Chr.15: 78565520-78595269 on Build GRCh38
Transcripts
NM_000745.3 NM_001307945.1 XM_005254142.3 XM_017021881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7273638 c.322T>A p.L108I Chr.15: 78588332-78588332 on Build GRCh38
965388 c.356A>G p.K119R Chr.15: 78588366-78588366 on Build GRCh38
4425831 c.364C>T p.R122C Chr.15: 78588374-78588374 on Build GRCh38
6392833 c.380C>A p.S127Y Chr.15: 78588390-78588390 on Build GRCh38
279894 c.375A>C p.S125S Chr.15: 78588385-78588385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12898919 Chr.15: 78588235-78588235 on Build GRCh38
rs778473235 Chr.15: 78588364-78588364 on Build GRCh38
rs759579004 Chr.15: 78588211-78588218 on Build GRCh38
rs761851654 Chr.15: 78588215-78588216 on Build GRCh38
rs947116397 Chr.15: 78588347-78588347 on Build GRCh38
rs910242839 Chr.15: 78588325-78588325 on Build GRCh38
rs371640436 Chr.15: 78588349-78588349 on Build GRCh38
rs72650654 Chr.15: 78588217-78588218 on Build GRCh38
rs565382338 Chr.15: 78588241-78588242 on Build GRCh38
rs754710874 Chr.15: 78588360-78588360 on Build GRCh38
rs932902057 Chr.15: 78588246-78588246 on Build GRCh38
rs767451621 Chr.15: 78588193-78588193 on Build GRCh38
rs752072464 Chr.15: 78588294-78588294 on Build GRCh38
rs949435276 Chr.15: 78588224-78588224 on Build GRCh38
rs199882720 Chr.15: 78588301-78588301 on Build GRCh38
rs201192025 Chr.15: 78588308-78588308 on Build GRCh38
rs56023835 Chr.15: 78588351-78588351 on Build GRCh38
rs774209829 Chr.15: 78588273-78588276 on Build GRCh38
rs952782136 Chr.15: 78588184-78588184 on Build GRCh38
rs201778201 Chr.15: 78588324-78588324 on Build GRCh38
rs977328308 Chr.15: 78588213-78588213 on Build GRCh38
rs376394314 Chr.15: 78588354-78588354 on Build GRCh38
rs961534169 Chr.15: 78588198-78588198 on Build GRCh38
rs531273490 Chr.15: 78588194-78588194 on Build GRCh38
rs926623440 Chr.15: 78588272-78588272 on Build GRCh38
rs745811241 Chr.15: 78588313-78588317 on Build GRCh38

View Full Product Details