Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40183527-40183796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1B
Entrez Gene ID
701
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase B
Gene Aliases
BUB1beta, BUBR1, Bub1A, MAD3L, MVA1, SSK1, hBUBR1
UniGene
Hs.513645
Gene Chromosome Location
Chr.15: 40161009-40221136 on Build GRCh38
Transcripts
NM_001211.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
159439 c.652G>C p.E218Q Chr.15: 40183784-40183784 on Build GRCh38
6142264 c.588C>G p.F196L Chr.15: 40183720-40183720 on Build GRCh38
3500795 c.613C>T p.L205L Chr.15: 40183745-40183745 on Build GRCh38
6448488 c.638G>T p.R213L Chr.15: 40183728-40183728 on Build GRCh38
3794095 c.659C>G p.S220C Chr.15: 40183791-40183791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775152576 Chr.15: 40183707-40183707 on Build GRCh38
rs779260067 Chr.15: 40183728-40183728 on Build GRCh38
rs756555949 Chr.15: 40183700-40183700 on Build GRCh38
rs759433961 Chr.15: 40183769-40183771 on Build GRCh38
rs888210381 Chr.15: 40183637-40183637 on Build GRCh38
rs767481744 Chr.15: 40183689-40183689 on Build GRCh38
rs759851663 Chr.15: 40183684-40183684 on Build GRCh38
rs778292065 Chr.15: 40183713-40183713 on Build GRCh38
rs746647687 Chr.15: 40183736-40183736 on Build GRCh38
rs768417752 Chr.15: 40183737-40183737 on Build GRCh38
rs751765984 Chr.15: 40183682-40183682 on Build GRCh38
rs981879563 Chr.15: 40183617-40183617 on Build GRCh38
rs747746384 Chr.15: 40183756-40183756 on Build GRCh38
rs148957803 Chr.15: 40183607-40183607 on Build GRCh38
rs530569397 Chr.15: 40183613-40183613 on Build GRCh38
rs368714094 Chr.15: 40183699-40183699 on Build GRCh38
rs1037175607 Chr.15: 40183719-40183719 on Build GRCh38
rs927300343 Chr.15: 40183618-40183618 on Build GRCh38
rs937462481 Chr.15: 40183661-40183661 on Build GRCh38
rs780541154 Chr.15: 40183743-40183743 on Build GRCh38
rs972504673 Chr.15: 40183569-40183569 on Build GRCh38
rs766311361 Chr.15: 40183681-40183681 on Build GRCh38
rs769213287 Chr.15: 40183768-40183768 on Build GRCh38
rs751339245 Chr.15: 40183761-40183763 on Build GRCh38
rs754371960 Chr.15: 40183725-40183725 on Build GRCh38
rs869263709 Chr.15: 40183629-40183629 on Build GRCh38

View Full Product Details