Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7732880-7733127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH2
Entrez Gene ID
146754
Gene Name
dynein axonemal heavy chain 2
Gene Aliases
DNAHC2, DNHD3
UniGene
Hs.367649
Gene Chromosome Location
Chr.17: 7717354-7833744 on Build GRCh38
Transcripts
NM_001303270.1 NM_020877.3 XM_011523663.1 XM_011523664.2 XM_011523666.1 XM_011523667.2 XM_011523670.2 XM_017024219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5853628 c.428G>A p.R143H Chr.17: 7733115-7733115 on Build GRCh38
1709741 c.427C>T p.R143C Chr.17: 7733114-7733114 on Build GRCh38
5853629 c.428G>A p.R143H Chr.17: 7733115-7733115 on Build GRCh38
4918149 c.403C>A p.Q135K Chr.17: 7733090-7733090 on Build GRCh38
77590 c.411G>C p.Q137H Chr.17: 7733098-7733098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898084478 Chr.17: 7733018-7733018 on Build GRCh38
rs763148087 Chr.17: 7733090-7733090 on Build GRCh38
rs770086862 Chr.17: 7733081-7733081 on Build GRCh38
rs148616573 Chr.17: 7732911-7732911 on Build GRCh38
rs375467657 Chr.17: 7733073-7733073 on Build GRCh38
rs113498896 Chr.17: 7732942-7732942 on Build GRCh38
rs995375122 Chr.17: 7733030-7733030 on Build GRCh38
rs778840302 Chr.17: 7733038-7733038 on Build GRCh38
rs1043943000 Chr.17: 7732957-7732957 on Build GRCh38
rs927992537 Chr.17: 7732908-7732908 on Build GRCh38
rs1038674483 Chr.17: 7733040-7733040 on Build GRCh38
rs935498866 Chr.17: 7732973-7732973 on Build GRCh38
rs187531102 Chr.17: 7732930-7732930 on Build GRCh38
rs990004153 Chr.17: 7732925-7732925 on Build GRCh38
rs906890046 Chr.17: 7732972-7732972 on Build GRCh38
rs370412978 Chr.17: 7733046-7733046 on Build GRCh38
rs1051207537 Chr.17: 7732981-7732981 on Build GRCh38
rs900150234 Chr.17: 7733048-7733048 on Build GRCh38
rs771839916 Chr.17: 7733060-7733060 on Build GRCh38
rs1027860879 Chr.17: 7733034-7733034 on Build GRCh38
rs539491504 Chr.17: 7733095-7733095 on Build GRCh38
rs557178381 Chr.17: 7733014-7733014 on Build GRCh38
rs146302909 Chr.17: 7733092-7733092 on Build GRCh38
rs568748840 Chr.17: 7733031-7733031 on Build GRCh38
rs924228556 Chr.17: 7732965-7732965 on Build GRCh38
rs550556480 Chr.17: 7732919-7732919 on Build GRCh38
rs774628888 Chr.17: 7733107-7733107 on Build GRCh38
rs192385531 Chr.17: 7733000-7733000 on Build GRCh38
rs745857145 Chr.17: 7733068-7733072 on Build GRCh38
rs575235290 Chr.17: 7733088-7733088 on Build GRCh38
rs546505764 Chr.17: 7732934-7732934 on Build GRCh38
rs939683862 Chr.17: 7732994-7732994 on Build GRCh38
rs1019006283 Chr.17: 7733052-7733052 on Build GRCh38
rs781348453 Chr.17: 7733068-7733068 on Build GRCh38

View Full Product Details