Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39333192-39333453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR8
Entrez Gene ID
1237
Gene Name
C-C motif chemokine receptor 8
Gene Aliases
CC-CKR-8, CCR-8, CDw198, CKRL1, CMKBR8, CMKBRL2, CY6, GPRCY6, TER1
UniGene
Hs.113222
Gene Chromosome Location
Chr.3: 39329706-39333680 on Build GRCh38
Transcripts
NM_005201.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3593918 c.872C>T p.T291I Chr.3: 39333203-39333203 on Build GRCh38
3593917 c.866C>T p.S289F Chr.3: 39333197-39333197 on Build GRCh38
5240774 c.931C>T p.L311F Chr.3: 39333262-39333262 on Build GRCh38
7091622 c.949_950insAAAGT p.C319fs*10 Chr.3: 39333280-39333281 on Build GRCh38
3727366 c.1046C>T p.S349F Chr.3: 39333377-39333377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760975188 Chr.3: 39333370-39333370 on Build GRCh38
rs775872835 Chr.3: 39333361-39333361 on Build GRCh38
rs764662305 Chr.3: 39333394-39333394 on Build GRCh38
rs749914662 Chr.3: 39333396-39333396 on Build GRCh38
rs771985291 Chr.3: 39333364-39333364 on Build GRCh38
rs774365244 Chr.3: 39333329-39333329 on Build GRCh38
rs151154954 Chr.3: 39333246-39333246 on Build GRCh38
rs765979465 Chr.3: 39333399-39333399 on Build GRCh38
rs140234572 Chr.3: 39333381-39333381 on Build GRCh38
rs200085677 Chr.3: 39333292-39333292 on Build GRCh38
rs756121637 Chr.3: 39333426-39333426 on Build GRCh38
rs764605073 Chr.3: 39333260-39333260 on Build GRCh38
rs902839284 Chr.3: 39333411-39333411 on Build GRCh38
rs754926627 Chr.3: 39333311-39333311 on Build GRCh38
rs748405129 Chr.3: 39333333-39333333 on Build GRCh38
rs201586986 Chr.3: 39333220-39333220 on Build GRCh38
rs146661016 Chr.3: 39333322-39333322 on Build GRCh38
rs200955972 Chr.3: 39333262-39333262 on Build GRCh38
rs764776284 Chr.3: 39333272-39333272 on Build GRCh38
rs201108728 Chr.3: 39333382-39333382 on Build GRCh38
rs946999491 Chr.3: 39333379-39333379 on Build GRCh38
rs771125610 Chr.3: 39333324-39333324 on Build GRCh38
rs772193443 Chr.3: 39333352-39333352 on Build GRCh38
rs748538869 Chr.3: 39333304-39333304 on Build GRCh38
rs757991973 Chr.3: 39333284-39333284 on Build GRCh38
rs566868115 Chr.3: 39333237-39333237 on Build GRCh38
rs748096777 Chr.3: 39333425-39333425 on Build GRCh38
rs746803713 Chr.3: 39333306-39333306 on Build GRCh38
rs376440421 Chr.3: 39333287-39333287 on Build GRCh38
rs150536198 Chr.3: 39333266-39333266 on Build GRCh38
rs781195359 Chr.3: 39333418-39333418 on Build GRCh38
rs758021269 Chr.3: 39333398-39333398 on Build GRCh38
rs754800684 Chr.3: 39333410-39333410 on Build GRCh38
rs767916882 Chr.3: 39333239-39333239 on Build GRCh38
rs764480741 Chr.3: 39333371-39333371 on Build GRCh38
rs781246660 Chr.3: 39333321-39333321 on Build GRCh38
rs769378560 Chr.3: 39333421-39333421 on Build GRCh38
rs750076765 Chr.3: 39333276-39333276 on Build GRCh38
rs915358783 Chr.3: 39333360-39333360 on Build GRCh38
rs751267843 Chr.3: 39333406-39333406 on Build GRCh38
rs981056176 Chr.3: 39333227-39333227 on Build GRCh38
rs761183744 Chr.3: 39333250-39333250 on Build GRCh38

View Full Product Details