Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41009967-41010200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2B6
Entrez Gene ID
1555
Gene Name
cytochrome P450 family 2 subfamily B member 6
Gene Aliases
CPB6, CYP2B, CYP2B7, CYP2B7P, CYPIIB6, EFVM, IIB1, P450
UniGene
Hs.1360
Gene Chromosome Location
Chr.19: 40991200-41018402 on Build GRCh38
Transcripts
NM_000767.4 XM_005258569.4 XM_006723050.3 XM_011526546.2 XM_011526547.2 XM_011526548.2 XM_011526549.2 XM_011526550.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2151063 c.850A>T p.S284C Chr.19: 41010021-41010021 on Build GRCh38
6219983 c.928G>A p.G310S Chr.19: 41010099-41010099 on Build GRCh38
474767 c.908C>T p.T303I Chr.19: 41010079-41010079 on Build GRCh38
7128417 c.870C>T p.L290L Chr.19: 41010041-41010041 on Build GRCh38
1393924 c.927C>T p.Y309Y Chr.19: 41010098-41010098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76471702 Chr.19: 41009991-41009991 on Build GRCh38
rs74869835 Chr.19: 41009993-41009993 on Build GRCh38
rs35468935 Chr.19: 41010104-41010104 on Build GRCh38
rs190463703 Chr.19: 41010027-41010027 on Build GRCh38
rs759397342 Chr.19: 41010036-41010036 on Build GRCh38
rs376762508 Chr.19: 41010153-41010153 on Build GRCh38
rs200503668 Chr.19: 41010177-41010177 on Build GRCh38
rs755545200 Chr.19: 41010163-41010163 on Build GRCh38
rs777943605 Chr.19: 41010149-41010150 on Build GRCh38
rs758553490 Chr.19: 41010158-41010158 on Build GRCh38
rs374642809 Chr.19: 41009989-41009989 on Build GRCh38
rs138650489 Chr.19: 41010146-41010147 on Build GRCh38
rs138594605 Chr.19: 41010046-41010046 on Build GRCh38
rs866444628 Chr.19: 41010015-41010015 on Build GRCh38
rs371440770 Chr.19: 41010005-41010005 on Build GRCh38
rs146617126 Chr.19: 41010028-41010028 on Build GRCh38
rs193922917 Chr.19: 41010108-41010108 on Build GRCh38
rs772533995 Chr.19: 41010012-41010012 on Build GRCh38
rs112848152 Chr.19: 41010047-41010047 on Build GRCh38
rs781543934 Chr.19: 41010172-41010172 on Build GRCh38
rs780229351 Chr.19: 41010096-41010096 on Build GRCh38
rs748862371 Chr.19: 41010174-41010174 on Build GRCh38
rs1024500804 Chr.19: 41010003-41010003 on Build GRCh38
rs34277950 Chr.19: 41010038-41010038 on Build GRCh38
rs770771771 Chr.19: 41010107-41010107 on Build GRCh38
rs373559488 Chr.19: 41010093-41010093 on Build GRCh38
rs143806947 Chr.19: 41010010-41010010 on Build GRCh38
rs141345415 Chr.19: 41010032-41010032 on Build GRCh38
rs535281317 Chr.19: 41009996-41009996 on Build GRCh38
rs776813682 Chr.19: 41010149-41010149 on Build GRCh38
rs763915587 Chr.19: 41010147-41010147 on Build GRCh38
rs768992518 Chr.19: 41010098-41010098 on Build GRCh38
rs139029625 Chr.19: 41010006-41010006 on Build GRCh38
rs774133160 Chr.19: 41010116-41010116 on Build GRCh38
rs763405622 Chr.19: 41010023-41010023 on Build GRCh38
rs760463009 Chr.19: 41010048-41010048 on Build GRCh38
rs757524845 Chr.19: 41010090-41010090 on Build GRCh38
rs747558979 Chr.19: 41010097-41010097 on Build GRCh38
rs777387885 Chr.19: 41010101-41010101 on Build GRCh38
rs745458475 Chr.19: 41010122-41010122 on Build GRCh38
rs751844933 Chr.19: 41010162-41010162 on Build GRCh38
rs200340276 Chr.19: 41010004-41010004 on Build GRCh38
rs750659675 Chr.19: 41010157-41010157 on Build GRCh38
rs34698757 Chr.19: 41010088-41010088 on Build GRCh38
rs758951550 Chr.19: 41010100-41010114 on Build GRCh38
rs765504158 Chr.19: 41010156-41010156 on Build GRCh38
rs778498584 Chr.19: 41010180-41010180 on Build GRCh38
rs760684934 Chr.19: 41010134-41010134 on Build GRCh38
rs775106445 Chr.19: 41010131-41010131 on Build GRCh38
rs374913591 Chr.19: 41010011-41010011 on Build GRCh38
rs754094512 Chr.19: 41009990-41009990 on Build GRCh38
rs771996518 Chr.19: 41010124-41010124 on Build GRCh38
rs758750908 Chr.19: 41010094-41010094 on Build GRCh38
rs763613336 Chr.19: 41010161-41010161 on Build GRCh38

View Full Product Details