Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231694386-231694637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DISC1
Entrez Gene ID
27185
Gene Name
disrupted in schizophrenia 1
Gene Aliases
C1orf136, SCZD9
UniGene
Hs.13318
Gene Chromosome Location
Chr.1: 231626815-232041272 on Build GRCh38
Transcripts
NM_001012957.1 NM_001012958.1 NM_001012959.1 NM_001164537.1 NM_001164538.1 NM_001164539.1 NM_001164540.1 NM_001164541.1 NM_001164542.1 NM_001164544.1 NM_001164545.1 NM_001164546.1 NM_001164547.1 NM_001164548.1 NM_001164549.1 NM_001164550.1 NM_001164551.1 NM_001164552.1 NM_001164553.1 NM_001164554.1 NM_001164555.1 NM_001164556.1 NM_018662.2 NR_028393.1 NR_028394.1 NR_028395.1 NR_028396.1 NR_028397.1 NR_028398.1 NR_028399.1 NR_028400.1

Gene Symbol
TSNAX-DISC1
Entrez Gene ID
100303453
Gene Name
TSNAX-DISC1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.13318
Gene Chromosome Location
Chr.1: 231528653-232041272 on Build GRCh38
Transcripts
NM_001012957.1 NM_001012958.1 NM_001012959.1 NM_001164537.1 NM_001164538.1 NM_001164539.1 NM_001164540.1 NM_001164541.1 NM_001164542.1 NM_001164544.1 NM_001164545.1 NM_001164546.1 NM_001164547.1 NM_001164548.1 NM_001164549.1 NM_001164550.1 NM_001164551.1 NM_001164552.1 NM_001164553.1 NM_001164554.1 NM_001164555.1 NM_001164556.1 NM_018662.2 NR_028393.1 NR_028394.1 NR_028395.1 NR_028396.1 NR_028397.1 NR_028398.1 NR_028399.1 NR_028400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1998581 c.717C>T p.C239C Chr.1: 231694475-231694475 on Build GRCh38
7216869 c.791G>A p.R264Q Chr.1: 231694549-231694549 on Build GRCh38
7106254 c.673G>A p.E225K Chr.1: 231694431-231694431 on Build GRCh38
1689965 c.671G>A p.G224E Chr.1: 231694429-231694429 on Build GRCh38
6541968 c.640C>T p.R214W Chr.1: 231694398-231694398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755631643 Chr.1: 231694593-231694593 on Build GRCh38
rs765577746 Chr.1: 231694579-231694579 on Build GRCh38
rs762099070 Chr.1: 231694487-231694487 on Build GRCh38
rs879001090 Chr.1: 231694597-231694597 on Build GRCh38
rs199530992 Chr.1: 231694468-231694468 on Build GRCh38
rs778749511 Chr.1: 231694613-231694613 on Build GRCh38
rs3738401 Chr.1: 231694549-231694549 on Build GRCh38
rs367543085 Chr.1: 231694526-231694526 on Build GRCh38
rs769513744 Chr.1: 231694422-231694422 on Build GRCh38
rs146439119 Chr.1: 231694426-231694426 on Build GRCh38
rs764251510 Chr.1: 231694480-231694480 on Build GRCh38
rs138228295 Chr.1: 231694598-231694598 on Build GRCh38
rs986232343 Chr.1: 231694461-231694461 on Build GRCh38
rs766759086 Chr.1: 231694588-231694588 on Build GRCh38
rs780719614 Chr.1: 231694518-231694518 on Build GRCh38
rs377484189 Chr.1: 231694535-231694535 on Build GRCh38
rs764811265 Chr.1: 231694527-231694529 on Build GRCh38
rs760628896 Chr.1: 231694565-231694565 on Build GRCh38
rs776775277 Chr.1: 231694572-231694572 on Build GRCh38
rs762545623 Chr.1: 231694485-231694485 on Build GRCh38
rs201249128 Chr.1: 231694456-231694456 on Build GRCh38
rs113312552 Chr.1: 231694517-231694517 on Build GRCh38
rs746525075 Chr.1: 231694417-231694417 on Build GRCh38
rs768870892 Chr.1: 231694571-231694571 on Build GRCh38
rs1055229412 Chr.1: 231694410-231694410 on Build GRCh38
rs373288445 Chr.1: 231694534-231694534 on Build GRCh38
rs750925617 Chr.1: 231694512-231694512 on Build GRCh38
rs750932225 Chr.1: 231694581-231694581 on Build GRCh38
rs775660207 Chr.1: 231694560-231694560 on Build GRCh38
rs750026954 Chr.1: 231694540-231694557 on Build GRCh38
rs765745403 Chr.1: 231694500-231694500 on Build GRCh38
rs148973353 Chr.1: 231694548-231694548 on Build GRCh38
rs757157671 Chr.1: 231694607-231694607 on Build GRCh38
rs201258158 Chr.1: 231694574-231694574 on Build GRCh38
rs867093901 Chr.1: 231694492-231694492 on Build GRCh38
rs763492083 Chr.1: 231694582-231694582 on Build GRCh38
rs997930434 Chr.1: 231694449-231694449 on Build GRCh38
rs759594286 Chr.1: 231694447-231694447 on Build GRCh38
rs201556843 Chr.1: 231694465-231694465 on Build GRCh38
rs752258922 Chr.1: 231694590-231694590 on Build GRCh38
rs773166710 Chr.1: 231694425-231694425 on Build GRCh38
rs777504676 Chr.1: 231694527-231694527 on Build GRCh38
rs754230811 Chr.1: 231694481-231694481 on Build GRCh38
rs754450096 Chr.1: 231694418-231694418 on Build GRCh38
rs779060951 Chr.1: 231694542-231694542 on Build GRCh38
rs373365825 Chr.1: 231694469-231694469 on Build GRCh38
rs747828145 Chr.1: 231694419-231694419 on Build GRCh38
rs367543086 Chr.1: 231694599-231694599 on Build GRCh38
rs375607284 Chr.1: 231694440-231694440 on Build GRCh38
rs199893176 Chr.1: 231694524-231694524 on Build GRCh38
rs767632505 Chr.1: 231694448-231694448 on Build GRCh38
rs141459782 Chr.1: 231694573-231694573 on Build GRCh38
rs753537068 Chr.1: 231694606-231694606 on Build GRCh38
rs149702548 Chr.1: 231694537-231694537 on Build GRCh38
rs755865070 Chr.1: 231694525-231694525 on Build GRCh38
rs774376929 Chr.1: 231694442-231694442 on Build GRCh38

View Full Product Details