Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4311735-4311972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSD1
Entrez Gene ID
79187
Gene Name
fibronectin type III and SPRY domain containing 1
Gene Aliases
GLFND, MIR1
UniGene
Hs.28144
Gene Chromosome Location
Chr.19: 4304594-4323846 on Build GRCh38
Transcripts
NM_024333.2 XM_005259644.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
292179 c.560G>T p.R187L Chr.19: 4311911-4311911 on Build GRCh38
3287937 c.560G>A p.R187H Chr.19: 4311911-4311911 on Build GRCh38
4780480 c.588C>A p.D196E Chr.19: 4311939-4311939 on Build GRCh38
1393211 c.611G>A p.R204Q Chr.19: 4311962-4311962 on Build GRCh38
6488295 c.549C>A p.T183T Chr.19: 4311900-4311900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774997278 Chr.19: 4311861-4311861 on Build GRCh38
rs773870728 Chr.19: 4311817-4311817 on Build GRCh38
rs745768424 Chr.19: 4311855-4311855 on Build GRCh38
rs371172774 Chr.19: 4311924-4311924 on Build GRCh38
rs147635723 Chr.19: 4311867-4311867 on Build GRCh38
rs762144186 Chr.19: 4311884-4311884 on Build GRCh38
rs755196317 Chr.19: 4311837-4311837 on Build GRCh38
rs747092389 Chr.19: 4311835-4311835 on Build GRCh38
rs950126714 Chr.19: 4311885-4311885 on Build GRCh38
rs765632467 Chr.19: 4311824-4311824 on Build GRCh38
rs376714727 Chr.19: 4311814-4311814 on Build GRCh38
rs893307833 Chr.19: 4311919-4311919 on Build GRCh38
rs764806397 Chr.19: 4311951-4311951 on Build GRCh38
rs773570628 Chr.19: 4311953-4311953 on Build GRCh38
rs377698035 Chr.19: 4311926-4311926 on Build GRCh38
rs771985733 Chr.19: 4311925-4311925 on Build GRCh38
rs767544615 Chr.19: 4311794-4311794 on Build GRCh38
rs748425183 Chr.19: 4311917-4311917 on Build GRCh38
rs774130281 Chr.19: 4311853-4311853 on Build GRCh38
rs763813323 Chr.19: 4311850-4311850 on Build GRCh38
rs1041552514 Chr.19: 4311815-4311815 on Build GRCh38
rs758878355 Chr.19: 4311905-4311905 on Build GRCh38
rs780147100 Chr.19: 4311834-4311834 on Build GRCh38
rs142307969 Chr.19: 4311910-4311910 on Build GRCh38
rs763733889 Chr.19: 4311863-4311863 on Build GRCh38
rs904886538 Chr.19: 4311928-4311928 on Build GRCh38
rs763738271 Chr.19: 4311813-4311813 on Build GRCh38
rs34155981 Chr.19: 4311918-4311918 on Build GRCh38
rs374955411 Chr.19: 4311798-4311798 on Build GRCh38
rs760189878 Chr.19: 4311862-4311862 on Build GRCh38
rs202097062 Chr.19: 4311831-4311831 on Build GRCh38
rs767099143 Chr.19: 4311796-4311796 on Build GRCh38
rs552864188 Chr.19: 4311789-4311789 on Build GRCh38
rs776308505 Chr.19: 4311864-4311864 on Build GRCh38
rs199532115 Chr.19: 4311944-4311944 on Build GRCh38
rs536463419 Chr.19: 4311785-4311785 on Build GRCh38
rs766812292 Chr.19: 4311908-4311908 on Build GRCh38
rs985087320 Chr.19: 4311830-4311830 on Build GRCh38
rs1003192870 Chr.19: 4311941-4311941 on Build GRCh38
rs749088245 Chr.19: 4311849-4311849 on Build GRCh38
rs545061368 Chr.19: 4311911-4311911 on Build GRCh38
rs34953789 Chr.19: 4311945-4311945 on Build GRCh38
rs567433795 Chr.19: 4311772-4311772 on Build GRCh38
rs760279832 Chr.19: 4311811-4311811 on Build GRCh38
rs774411323 Chr.19: 4311792-4311792 on Build GRCh38
rs745655176 Chr.19: 4311923-4311923 on Build GRCh38
rs940483157 Chr.19: 4311844-4311844 on Build GRCh38
rs750787554 Chr.19: 4311900-4311900 on Build GRCh38
rs771934110 Chr.19: 4311856-4311856 on Build GRCh38
rs757686442 Chr.19: 4311820-4311820 on Build GRCh38
rs781619574 Chr.19: 4311847-4311847 on Build GRCh38
rs373393339 Chr.19: 4311780-4311781 on Build GRCh38
rs781531917 Chr.19: 4311913-4311913 on Build GRCh38
rs770976294 Chr.19: 4311791-4311791 on Build GRCh38
rs750988761 Chr.19: 4311825-4311825 on Build GRCh38
rs750526066 Chr.19: 4311892-4311893 on Build GRCh38
rs544256843 Chr.19: 4311949-4311949 on Build GRCh38
rs373678270 Chr.19: 4311939-4311939 on Build GRCh38
rs765588543 Chr.19: 4311895-4311895 on Build GRCh38

View Full Product Details