Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15644444-15644669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTD
Entrez Gene ID
686
Gene Name
biotinidase
Gene Aliases
-
UniGene
Hs.444197
Gene Chromosome Location
Chr.3: 15601352-15653709 on Build GRCh38
Transcripts
XM_006713314.3 XM_011534041.2 XM_017007088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5549120 c.710C>T p.A237V Chr.3: 15644566-15644566 on Build GRCh38
5556430 c.795T>C p.H265H Chr.3: 15644651-15644651 on Build GRCh38
6613464 c.701delC p.F236fs*28 Chr.3: 15644557-15644557 on Build GRCh38
6613466 c.632G>A p.R211H Chr.3: 15644488-15644488 on Build GRCh38
3005349 c.712G>T p.G238C Chr.3: 15644568-15644568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145388314 Chr.3: 15644501-15644501 on Build GRCh38
rs397514378 Chr.3: 15644508-15644508 on Build GRCh38
rs946525644 Chr.3: 15644486-15644486 on Build GRCh38
rs773669239 Chr.3: 15644494-15644494 on Build GRCh38
rs372844636 Chr.3: 15644487-15644487 on Build GRCh38
rs746554189 Chr.3: 15644470-15644470 on Build GRCh38
rs112195009 Chr.3: 15644488-15644488 on Build GRCh38
rs398123140 Chr.3: 15644598-15644598 on Build GRCh38
rs746493935 Chr.3: 15644548-15644548 on Build GRCh38
rs140769832 Chr.3: 15644561-15644561 on Build GRCh38
rs763366957 Chr.3: 15644574-15644574 on Build GRCh38
rs28934601 Chr.3: 15644611-15644611 on Build GRCh38
rs777224767 Chr.3: 15644588-15644588 on Build GRCh38
rs1056525743 Chr.3: 15644560-15644560 on Build GRCh38
rs190386869 Chr.3: 15644499-15644499 on Build GRCh38
rs369102875 Chr.3: 15644481-15644481 on Build GRCh38
rs752930467 Chr.3: 15644594-15644594 on Build GRCh38
rs758306005 Chr.3: 15644612-15644612 on Build GRCh38
rs587783004 Chr.3: 15644539-15644539 on Build GRCh38
rs398123139 Chr.3: 15644482-15644482 on Build GRCh38
rs145303558 Chr.3: 15644526-15644526 on Build GRCh38
rs777470168 Chr.3: 15644645-15644645 on Build GRCh38
rs767750851 Chr.3: 15644507-15644507 on Build GRCh38
rs780874850 Chr.3: 15644493-15644493 on Build GRCh38
rs771466219 Chr.3: 15644580-15644580 on Build GRCh38
rs779919717 Chr.3: 15644540-15644540 on Build GRCh38
rs368913302 Chr.3: 15644635-15644635 on Build GRCh38
rs587783005 Chr.3: 15644557-15644557 on Build GRCh38
rs397507175 Chr.3: 15644590-15644590 on Build GRCh38
rs397514381 Chr.3: 15644565-15644565 on Build GRCh38
rs397507174 Chr.3: 15644485-15644485 on Build GRCh38
rs1044823012 Chr.3: 15644495-15644495 on Build GRCh38
rs757071809 Chr.3: 15644516-15644516 on Build GRCh38
rs767518580 Chr.3: 15644591-15644591 on Build GRCh38
rs142418812 Chr.3: 15644519-15644519 on Build GRCh38
rs1003975438 Chr.3: 15644532-15644532 on Build GRCh38
rs768097543 Chr.3: 15644551-15644551 on Build GRCh38
rs397514380 Chr.3: 15644538-15644538 on Build GRCh38
rs780017297 Chr.3: 15644616-15644616 on Build GRCh38
rs372687866 Chr.3: 15644639-15644639 on Build GRCh38
rs144901367 Chr.3: 15644569-15644569 on Build GRCh38
rs397514379 Chr.3: 15644510-15644510 on Build GRCh38
rs397514382 Chr.3: 15644599-15644599 on Build GRCh38
rs397514377 Chr.3: 15644497-15644497 on Build GRCh38
rs200337373 Chr.3: 15644520-15644520 on Build GRCh38
rs769423080 Chr.3: 15644643-15644643 on Build GRCh38
rs1035402421 Chr.3: 15644537-15644537 on Build GRCh38
rs397514384 Chr.3: 15644620-15644620 on Build GRCh38
rs397514383 Chr.3: 15644613-15644613 on Build GRCh38
rs770075741 Chr.3: 15644562-15644562 on Build GRCh38
rs367609937 Chr.3: 15644632-15644632 on Build GRCh38
rs765071585 Chr.3: 15644605-15644605 on Build GRCh38
rs397514438 Chr.3: 15644614-15644614 on Build GRCh38
rs774761127 Chr.3: 15644585-15644585 on Build GRCh38

View Full Product Details