Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92349280-92349505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARCH5
Entrez Gene ID
54708
Gene Name
membrane associated ring-CH-type finger 5
Gene Aliases
MARCH-V, MITOL, RNF153
UniGene
Hs.573490
Gene Chromosome Location
Chr.10: 92291163-92353964 on Build GRCh38
Transcripts
NM_017824.4 XM_011539886.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6708245 c.400A>G p.M134V Chr.10: 92349379-92349379 on Build GRCh38
4776361 c.479G>A p.R160H Chr.10: 92349458-92349458 on Build GRCh38
921457 c.390T>A p.G130G Chr.10: 92349369-92349369 on Build GRCh38
428132 c.508C>T p.R170C Chr.10: 92349487-92349487 on Build GRCh38
6708243 c.509G>A p.R170H Chr.10: 92349488-92349488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988758779 Chr.10: 92349433-92349433 on Build GRCh38
rs765165369 Chr.10: 92349449-92349449 on Build GRCh38
rs761902052 Chr.10: 92349410-92349410 on Build GRCh38
rs764021315 Chr.10: 92349399-92349399 on Build GRCh38
rs956416825 Chr.10: 92349424-92349424 on Build GRCh38
rs749904400 Chr.10: 92349472-92349472 on Build GRCh38
rs370134787 Chr.10: 92349466-92349466 on Build GRCh38
rs1035069324 Chr.10: 92349414-92349414 on Build GRCh38
rs375467796 Chr.10: 92349340-92349340 on Build GRCh38
rs747355836 Chr.10: 92349328-92349328 on Build GRCh38
rs367936216 Chr.10: 92349324-92349324 on Build GRCh38
rs778054509 Chr.10: 92349309-92349309 on Build GRCh38
rs775736039 Chr.10: 92349337-92349337 on Build GRCh38
rs372532174 Chr.10: 92349336-92349336 on Build GRCh38
rs776564011 Chr.10: 92349405-92349405 on Build GRCh38
rs867130938 Chr.10: 92349481-92349481 on Build GRCh38

View Full Product Details