Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190683547-190683788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAB1
Entrez Gene ID
4664
Gene Name
NGFI-A binding protein 1
Gene Aliases
-
UniGene
Hs.107474
Gene Chromosome Location
Chr.2: 190648860-190692766 on Build GRCh38
Transcripts
NM_001321312.1 NM_001321313.1 NM_001321314.1 NM_001321315.1 NM_005966.3 XM_005246582.1 XM_005246583.1 XM_005246586.1 XM_005246587.1 XM_011511219.2 XM_017004170.1 XM_017004171.1 XM_017004172.1 XM_017004173.1 XM_017004174.1 XM_017004175.1 XM_017004176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4921407 c.1010G>T p.G337V Chr.2: 190683742-190683742 on Build GRCh38
1631671 c.1006-3C>T p.? Chr.2: 190683735-190683735 on Build GRCh38
5395820 c.1011G>A p.G337G Chr.2: 190683743-190683743 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs539589193 Chr.2: 190683681-190683681 on Build GRCh38
rs372594920 Chr.2: 190683767-190683767 on Build GRCh38
rs953802346 Chr.2: 190683593-190683593 on Build GRCh38
rs772323667 Chr.2: 190683711-190683711 on Build GRCh38
rs753747345 Chr.2: 190683737-190683737 on Build GRCh38
rs756135415 Chr.2: 190683614-190683614 on Build GRCh38
rs766791888 Chr.2: 190683727-190683727 on Build GRCh38
rs763671335 Chr.2: 190683697-190683697 on Build GRCh38
rs762055969 Chr.2: 190683702-190683702 on Build GRCh38
rs970026272 Chr.2: 190683722-190683722 on Build GRCh38
rs763718186 Chr.2: 190683763-190683763 on Build GRCh38
rs918886816 Chr.2: 190683709-190683709 on Build GRCh38
rs555835810 Chr.2: 190683644-190683644 on Build GRCh38
rs1056477193 Chr.2: 190683732-190683732 on Build GRCh38
rs377093347 Chr.2: 190683686-190683686 on Build GRCh38
rs777200201 Chr.2: 190683743-190683743 on Build GRCh38
rs910037401 Chr.2: 190683675-190683675 on Build GRCh38
rs762494622 Chr.2: 190683752-190683752 on Build GRCh38
rs150828744 Chr.2: 190683715-190683715 on Build GRCh38

View Full Product Details