Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 36519930-36520191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKX2-1
Entrez Gene ID
7080
Gene Name
NK2 homeobox 1
Gene Aliases
BCH, BHC, NK-2, NKX2.1, NKX2A, NMTC1, T/EBP, TEBP, TITF1, TTF-1, TTF1
UniGene
Hs.94367
Gene Chromosome Location
Chr.14: 36516397-36520225 on Build GRCh38
Transcripts
NM_001079668.2 NR_103710.1

Gene Symbol
NKX2-1-AS1
Entrez Gene ID
100506237
Gene Name
NKX2-1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.497090
Gene Chromosome Location
Chr.14: 36519278-36523016 on Build GRCh38
Transcripts
NM_001079668.2 NR_103710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6931375 c.41C>T p.A14V Chr.14: 36520089-36520089 on Build GRCh38
6970002 c.47C>A p.A16E Chr.14: 36520083-36520083 on Build GRCh38
6968946 c.10G>A p.G4R Chr.14: 36520120-36520120 on Build GRCh38
6970301 c.28C>T p.R10W Chr.14: 36520102-36520102 on Build GRCh38
6235482 c.8C>A p.S3Y Chr.14: 36520122-36520122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770066579 Chr.14: 36520059-36520059 on Build GRCh38
rs771149835 Chr.14: 36520074-36520074 on Build GRCh38
rs946195342 Chr.14: 36519994-36519994 on Build GRCh38
rs768690786 Chr.14: 36520132-36520132 on Build GRCh38
rs1045985330 Chr.14: 36520001-36520001 on Build GRCh38
rs201979429 Chr.14: 36520101-36520101 on Build GRCh38
rs372925758 Chr.14: 36520120-36520120 on Build GRCh38
rs529122651 Chr.14: 36519984-36519984 on Build GRCh38
rs555458391 Chr.14: 36519998-36519998 on Build GRCh38
rs759606678 Chr.14: 36520141-36520141 on Build GRCh38
rs749986225 Chr.14: 36520122-36520122 on Build GRCh38
rs771715414 Chr.14: 36520036-36520036 on Build GRCh38
rs918077408 Chr.14: 36520153-36520153 on Build GRCh38
rs774393926 Chr.14: 36520082-36520082 on Build GRCh38
rs758109939 Chr.14: 36520174-36520174 on Build GRCh38
rs763645944 Chr.14: 36520106-36520106 on Build GRCh38
rs778038773 Chr.14: 36520135-36520135 on Build GRCh38
rs755475569 Chr.14: 36520003-36520003 on Build GRCh38
rs1005311801 Chr.14: 36520112-36520112 on Build GRCh38
rs375793163 Chr.14: 36520128-36520128 on Build GRCh38
rs749002422 Chr.14: 36520023-36520023 on Build GRCh38
rs764617662 Chr.14: 36520155-36520155 on Build GRCh38
rs537795327 Chr.14: 36520162-36520162 on Build GRCh38
rs764586138 Chr.14: 36520168-36520168 on Build GRCh38
rs965527168 Chr.14: 36519988-36519988 on Build GRCh38
rs761213689 Chr.14: 36520149-36520149 on Build GRCh38
rs759963311 Chr.14: 36520094-36520094 on Build GRCh38
rs955469214 Chr.14: 36520161-36520161 on Build GRCh38
rs566529892 Chr.14: 36519983-36519983 on Build GRCh38
rs755630277 Chr.14: 36520018-36520018 on Build GRCh38
rs767808984 Chr.14: 36520098-36520098 on Build GRCh38
rs747000470 Chr.14: 36520129-36520129 on Build GRCh38
rs1045150494 Chr.14: 36520165-36520165 on Build GRCh38
rs990160382 Chr.14: 36519979-36519979 on Build GRCh38
rs1028441864 Chr.14: 36520035-36520035 on Build GRCh38
rs753150454 Chr.14: 36520015-36520015 on Build GRCh38
rs367897299 Chr.14: 36520148-36520148 on Build GRCh38
rs777264286 Chr.14: 36520021-36520021 on Build GRCh38
rs756914430 Chr.14: 36520024-36520024 on Build GRCh38
rs193043968 Chr.14: 36519978-36519978 on Build GRCh38
rs778614432 Chr.14: 36520034-36520034 on Build GRCh38
rs750156580 Chr.14: 36520170-36520170 on Build GRCh38
rs753273268 Chr.14: 36520099-36520099 on Build GRCh38
rs749414192 Chr.14: 36520137-36520137 on Build GRCh38
rs746875657 Chr.14: 36520051-36520051 on Build GRCh38
rs778651793 Chr.14: 36520121-36520121 on Build GRCh38
rs767485199 Chr.14: 36520134-36520135 on Build GRCh38
rs772486588 Chr.14: 36520143-36520143 on Build GRCh38
rs1009827872 Chr.14: 36519971-36519971 on Build GRCh38
rs753318699 Chr.14: 36520109-36520109 on Build GRCh38
rs754252302 Chr.14: 36520157-36520157 on Build GRCh38
rs774642011 Chr.14: 36520140-36520140 on Build GRCh38
rs568956283 Chr.14: 36520138-36520138 on Build GRCh38
rs780154530 Chr.14: 36520043-36520043 on Build GRCh38
rs763132470 Chr.14: 36520073-36520073 on Build GRCh38
rs768098744 Chr.14: 36520004-36520004 on Build GRCh38
rs773410433 Chr.14: 36520063-36520063 on Build GRCh38
rs952355601 Chr.14: 36520042-36520042 on Build GRCh38
rs530304913 Chr.14: 36520131-36520131 on Build GRCh38

View Full Product Details