Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50508393-50508644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAPH2
Entrez Gene ID
29781
Gene Name
non-SMC condensin II complex subunit H2
Gene Aliases
CAPH2
UniGene
Hs.730607
Gene Chromosome Location
Chr.22: 50508216-50524780 on Build GRCh38
Transcripts
NM_001185011.1 NM_014551.4 NM_152299.3 XM_005261912.4 XM_011530685.2 XM_017028793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4554061 c.60G>A p.K20K Chr.22: 50508397-50508397 on Build GRCh38
1566385 c.91G>T p.G31C Chr.22: 50508428-50508428 on Build GRCh38
1417146 c.60G>T p.K20N Chr.22: 50508397-50508397 on Build GRCh38
7379081 c.106G>A p.E36K Chr.22: 50508443-50508443 on Build GRCh38
6367253 c.80C>T p.A27V Chr.22: 50508417-50508417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746430594 Chr.22: 50508460-50508460 on Build GRCh38
rs568440240 Chr.22: 50508458-50508458 on Build GRCh38
rs1019697065 Chr.22: 50508439-50508439 on Build GRCh38
rs939677825 Chr.22: 50508593-50508593 on Build GRCh38
rs907524365 Chr.22: 50508416-50508416 on Build GRCh38
rs977591621 Chr.22: 50508486-50508486 on Build GRCh38
rs917016236 Chr.22: 50508615-50508615 on Build GRCh38
rs529402637 Chr.22: 50508466-50508466 on Build GRCh38
rs770520702 Chr.22: 50508456-50508456 on Build GRCh38
rs1028127642 Chr.22: 50508531-50508531 on Build GRCh38
rs550773675 Chr.22: 50508516-50508516 on Build GRCh38
rs775983277 Chr.22: 50508494-50508494 on Build GRCh38
rs997076243 Chr.22: 50508611-50508611 on Build GRCh38
rs928866908 Chr.22: 50508499-50508499 on Build GRCh38
rs992562162 Chr.22: 50508609-50508609 on Build GRCh38
rs917116836 Chr.22: 50508509-50508509 on Build GRCh38
rs141530541 Chr.22: 50508527-50508527 on Build GRCh38
rs956727985 Chr.22: 50508418-50508418 on Build GRCh38
rs908239467 Chr.22: 50508568-50508568 on Build GRCh38
rs131823 Chr.22: 50508536-50508536 on Build GRCh38
rs967522752 Chr.22: 50508474-50508474 on Build GRCh38
rs935984478 Chr.22: 50508547-50508547 on Build GRCh38
rs960342670 Chr.22: 50508505-50508505 on Build GRCh38
rs965230234 Chr.22: 50508518-50508518 on Build GRCh38
rs748807378 Chr.22: 50508421-50508421 on Build GRCh38
rs920489260 Chr.22: 50508468-50508468 on Build GRCh38
rs1054879742 Chr.22: 50508581-50508581 on Build GRCh38
rs765526742 Chr.22: 50508495-50508522 on Build GRCh38
rs952742254 Chr.22: 50508532-50508532 on Build GRCh38

View Full Product Details