Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63508420-63508649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGALS12
Entrez Gene ID
85329
Gene Name
galectin 12
Gene Aliases
GAL12, GRIP1
UniGene
Hs.502774
Gene Chromosome Location
Chr.11: 63506052-63516774 on Build GRCh38
Transcripts
NM_001142535.1 NM_001142536.1 NM_001142537.1 NM_001142538.1 NM_033101.3 XM_017018458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3451278 c.209C>T p.P70L Chr.11: 63508626-63508626 on Build GRCh38
4531956 c.183G>T p.K61N Chr.11: 63508600-63508600 on Build GRCh38
429495 c.175G>A p.A59T Chr.11: 63508592-63508592 on Build GRCh38
2162469 c.156G>A p.T52T Chr.11: 63508573-63508573 on Build GRCh38
3359288 c.165A>G p.G55G Chr.11: 63508582-63508582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970091614 Chr.11: 63508593-63508593 on Build GRCh38
rs775850965 Chr.11: 63508535-63508535 on Build GRCh38
rs775666434 Chr.11: 63508611-63508611 on Build GRCh38
rs972329836 Chr.11: 63508510-63508510 on Build GRCh38
rs144707121 Chr.11: 63508588-63508588 on Build GRCh38
rs771301477 Chr.11: 63508508-63508508 on Build GRCh38
rs966430152 Chr.11: 63508607-63508607 on Build GRCh38
rs111264048 Chr.11: 63508542-63508542 on Build GRCh38
rs746816515 Chr.11: 63508587-63508587 on Build GRCh38
rs577461705 Chr.11: 63508616-63508616 on Build GRCh38
rs893431550 Chr.11: 63508579-63508579 on Build GRCh38
rs749686664 Chr.11: 63508623-63508624 on Build GRCh38
rs752546690 Chr.11: 63508570-63508570 on Build GRCh38
rs915922172 Chr.11: 63508592-63508592 on Build GRCh38
rs753590832 Chr.11: 63508560-63508560 on Build GRCh38
rs781564834 Chr.11: 63508594-63508594 on Build GRCh38
rs182560081 Chr.11: 63508554-63508554 on Build GRCh38
rs376328731 Chr.11: 63508534-63508534 on Build GRCh38
rs763551709 Chr.11: 63508540-63508540 on Build GRCh38
rs776798608 Chr.11: 63508523-63508523 on Build GRCh38
rs769727603 Chr.11: 63508600-63508600 on Build GRCh38
rs746162900 Chr.11: 63508598-63508598 on Build GRCh38
rs754432350 Chr.11: 63508566-63508566 on Build GRCh38
rs1051933656 Chr.11: 63508526-63508526 on Build GRCh38
rs898005837 Chr.11: 63508481-63508481 on Build GRCh38
rs144263194 Chr.11: 63508569-63508569 on Build GRCh38
rs760259326 Chr.11: 63508556-63508556 on Build GRCh38
rs528636833 Chr.11: 63508585-63508585 on Build GRCh38
rs745941742 Chr.11: 63508529-63508529 on Build GRCh38
rs143717054 Chr.11: 63508487-63508487 on Build GRCh38
rs747194523 Chr.11: 63508494-63508494 on Build GRCh38

View Full Product Details