Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44800381-44800630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLK3
Entrez Gene ID
1263
Gene Name
polo like kinase 3
Gene Aliases
CNK, FNK, PLK-3, PRK
UniGene
Hs.632415
Gene Chromosome Location
Chr.1: 44799952-44805995 on Build GRCh38
Transcripts
NM_004073.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6045207 c.71C>T p.A24V Chr.1: 44800534-44800534 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs960214996 Chr.1: 44800413-44800413 on Build GRCh38
rs552804138 Chr.1: 44800423-44800423 on Build GRCh38
rs566229457 Chr.1: 44800501-44800501 on Build GRCh38
rs773595141 Chr.1: 44800600-44800600 on Build GRCh38
rs568357012 Chr.1: 44800574-44800574 on Build GRCh38
rs1008075120 Chr.1: 44800516-44800516 on Build GRCh38
rs890514336 Chr.1: 44800511-44800511 on Build GRCh38
rs756897955 Chr.1: 44800440-44800440 on Build GRCh38
rs879668424 Chr.1: 44800455-44800455 on Build GRCh38
rs554740111 Chr.1: 44800542-44800542 on Build GRCh38
rs968361297 Chr.1: 44800572-44800572 on Build GRCh38
rs535015888 Chr.1: 44800539-44800539 on Build GRCh38
rs770104520 Chr.1: 44800597-44800597 on Build GRCh38
rs996766098 Chr.1: 44800563-44800563 on Build GRCh38
rs767963385 Chr.1: 44800608-44800608 on Build GRCh38
rs778305430 Chr.1: 44800491-44800491 on Build GRCh38
rs769289342 Chr.1: 44800564-44800564 on Build GRCh38
rs993379752 Chr.1: 44800417-44800417 on Build GRCh38
rs929413783 Chr.1: 44800486-44800486 on Build GRCh38
rs372090200 Chr.1: 44800595-44800595 on Build GRCh38
rs1041292114 Chr.1: 44800547-44800547 on Build GRCh38
rs757088659 Chr.1: 44800499-44800499 on Build GRCh38
rs1040866447 Chr.1: 44800518-44800518 on Build GRCh38
rs112086768 Chr.1: 44800570-44800570 on Build GRCh38
rs924801697 Chr.1: 44800402-44800402 on Build GRCh38
rs916174443 Chr.1: 44800428-44800428 on Build GRCh38
rs948939006 Chr.1: 44800429-44800429 on Build GRCh38

View Full Product Details