Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48944119-48944380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUD5
Entrez Gene ID
55593
Gene Name
OTU deubiquitinase 5
Gene Aliases
DUBA
UniGene
Hs.496098
Gene Chromosome Location
Chr.X: 48922024-48958386 on Build GRCh38
Transcripts
NM_001136157.1 NM_001136158.1 NM_001136159.1 NM_017602.3 XM_005272622.3 XM_005278039.3 XM_005278042.4 XM_006724537.2 XM_011543932.1 XM_011543934.2 XM_017029628.1 XM_017029629.1 XM_017029630.1

SNP Detail

SNP ID Chromosome Location
rs782352138 Chr.X: 48944228-48944228 on Build GRCh38
rs782002385 Chr.X: 48944221-48944221 on Build GRCh38
rs782719600 Chr.X: 48944297-48944297 on Build GRCh38
rs782631321 Chr.X: 48944148-48944148 on Build GRCh38
rs933397226 Chr.X: 48944344-48944344 on Build GRCh38
rs149616505 Chr.X: 48944263-48944263 on Build GRCh38
rs782307788 Chr.X: 48944161-48944161 on Build GRCh38
rs781836867 Chr.X: 48944333-48944333 on Build GRCh38
rs782636234 Chr.X: 48944321-48944321 on Build GRCh38
rs782666250 Chr.X: 48944211-48944211 on Build GRCh38
rs200995116 Chr.X: 48944257-48944257 on Build GRCh38
rs782191211 Chr.X: 48944156-48944156 on Build GRCh38
rs782206772 Chr.X: 48944222-48944222 on Build GRCh38
rs782463846 Chr.X: 48944194-48944194 on Build GRCh38
rs782025856 Chr.X: 48944172-48944172 on Build GRCh38
rs782586700 Chr.X: 48944278-48944278 on Build GRCh38
rs782779725 Chr.X: 48944313-48944313 on Build GRCh38
rs781921785 Chr.X: 48944233-48944233 on Build GRCh38
rs781840824 Chr.X: 48944277-48944277 on Build GRCh38
rs782788304 Chr.X: 48944242-48944242 on Build GRCh38
rs375404510 Chr.X: 48944296-48944296 on Build GRCh38
rs781936828 Chr.X: 48944296-48944296 on Build GRCh38
rs781894635 Chr.X: 48944312-48944312 on Build GRCh38
rs782067268 Chr.X: 48944238-48944238 on Build GRCh38
rs782027513 Chr.X: 48944255-48944255 on Build GRCh38

View Full Product Details