Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55027511-55027766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GP6
Entrez Gene ID
51206
Gene Name
glycoprotein VI platelet
Gene Aliases
BDPLT11, GPIV, GPVI
UniGene
Hs.661752
Gene Chromosome Location
Chr.19: 55013705-55038264 on Build GRCh38
Transcripts
NM_001083899.2 NM_001256017.2 NM_016363.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1396470 c.576A>G p.S192S Chr.19: 55027612-55027612 on Build GRCh38
5814601 c.425A>T p.Q142L Chr.19: 55027763-55027763 on Build GRCh38
6582443 c.535C>T p.R179* Chr.19: 55027653-55027653 on Build GRCh38
4931857 c.573G>T p.W191C Chr.19: 55027615-55027615 on Build GRCh38
7304501 c.591C>A p.P197P Chr.19: 55027597-55027597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779552982 Chr.19: 55027602-55027602 on Build GRCh38
rs929541571 Chr.19: 55027550-55027550 on Build GRCh38
rs201652983 Chr.19: 55027621-55027621 on Build GRCh38
rs754684731 Chr.19: 55027713-55027713 on Build GRCh38
rs777476310 Chr.19: 55027683-55027683 on Build GRCh38
rs778314910 Chr.19: 55027598-55027598 on Build GRCh38
rs748027211 Chr.19: 55027675-55027675 on Build GRCh38
rs200525940 Chr.19: 55027604-55027604 on Build GRCh38
rs892089 Chr.19: 55027693-55027693 on Build GRCh38
rs760915271 Chr.19: 55027571-55027571 on Build GRCh38
rs1654425 Chr.19: 55027612-55027612 on Build GRCh38
rs897971250 Chr.19: 55027692-55027692 on Build GRCh38
rs746018547 Chr.19: 55027733-55027733 on Build GRCh38
rs369568059 Chr.19: 55027635-55027635 on Build GRCh38
rs751979972 Chr.19: 55027652-55027652 on Build GRCh38
rs766027583 Chr.19: 55027708-55027708 on Build GRCh38
rs755737487 Chr.19: 55027599-55027599 on Build GRCh38
rs370855777 Chr.19: 55027610-55027610 on Build GRCh38
rs969065691 Chr.19: 55027714-55027714 on Build GRCh38
rs911861220 Chr.19: 55027576-55027576 on Build GRCh38
rs746844654 Chr.19: 55027687-55027687 on Build GRCh38
rs755444298 Chr.19: 55027577-55027577 on Build GRCh38
rs746798268 Chr.19: 55027726-55027726 on Build GRCh38
rs943976459 Chr.19: 55027561-55027561 on Build GRCh38
rs766430706 Chr.19: 55027574-55027574 on Build GRCh38
rs373910879 Chr.19: 55027575-55027575 on Build GRCh38
rs571307915 Chr.19: 55027738-55027738 on Build GRCh38
rs900564009 Chr.19: 55027608-55027608 on Build GRCh38
rs367907292 Chr.19: 55027663-55027663 on Build GRCh38
rs558115658 Chr.19: 55027538-55027538 on Build GRCh38
rs759834481 Chr.19: 55027697-55027697 on Build GRCh38
rs778421895 Chr.19: 55027674-55027674 on Build GRCh38
rs5030705 Chr.19: 55027681-55027681 on Build GRCh38
rs768628583 Chr.19: 55027603-55027603 on Build GRCh38
rs753098085 Chr.19: 55027587-55027587 on Build GRCh38
rs781062868 Chr.19: 55027732-55027732 on Build GRCh38
rs768485791 Chr.19: 55027533-55027533 on Build GRCh38
rs41301959 Chr.19: 55027615-55027615 on Build GRCh38
rs754483221 Chr.19: 55027672-55027672 on Build GRCh38
rs776766695 Chr.19: 55027616-55027616 on Build GRCh38
rs886043669 Chr.19: 55027709-55027709 on Build GRCh38
rs758328364 Chr.19: 55027717-55027717 on Build GRCh38
rs1033464335 Chr.19: 55027723-55027723 on Build GRCh38
rs901882655 Chr.19: 55027584-55027584 on Build GRCh38
rs758874083 Chr.19: 55027588-55027588 on Build GRCh38
rs771170249 Chr.19: 55027691-55027691 on Build GRCh38
rs544932180 Chr.19: 55027549-55027549 on Build GRCh38
rs767700826 Chr.19: 55027554-55027554 on Build GRCh38
rs777746272 Chr.19: 55027719-55027719 on Build GRCh38
rs201166741 Chr.19: 55027741-55027741 on Build GRCh38
rs763485474 Chr.19: 55027702-55027702 on Build GRCh38
rs756962244 Chr.19: 55027727-55027727 on Build GRCh38
rs387906919 Chr.19: 55027664-55027664 on Build GRCh38
rs746720985 Chr.19: 55027575-55027575 on Build GRCh38
rs546902436 Chr.19: 55027633-55027633 on Build GRCh38
rs771165550 Chr.19: 55027703-55027704 on Build GRCh38
rs892090 Chr.19: 55027704-55027704 on Build GRCh38
rs755790003 Chr.19: 55027662-55027662 on Build GRCh38
rs753396840 Chr.19: 55027711-55027711 on Build GRCh38
rs762130319 Chr.19: 55027705-55027705 on Build GRCh38
rs534339924 Chr.19: 55027613-55027613 on Build GRCh38
rs931967435 Chr.19: 55027590-55027590 on Build GRCh38
rs773459051 Chr.19: 55027560-55027560 on Build GRCh38
rs41300084 Chr.19: 55027620-55027620 on Build GRCh38
rs764497463 Chr.19: 55027649-55027649 on Build GRCh38
rs765744190 Chr.19: 55027623-55027623 on Build GRCh38
rs769947136 Chr.19: 55027700-55027700 on Build GRCh38
rs778339783 Chr.19: 55027716-55027716 on Build GRCh38
rs753216495 Chr.19: 55027625-55027625 on Build GRCh38
rs200041269 Chr.19: 55027536-55027536 on Build GRCh38
rs775726835 Chr.19: 55027701-55027701 on Build GRCh38

View Full Product Details