Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 131446318-131446543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRRC2B
Entrez Gene ID
84726
Gene Name
proline rich coiled-coil 2B
Gene Aliases
BAT2L, BAT2L1, KIAA0515, LQFBS-1
UniGene
Hs.743955
Gene Chromosome Location
Chr.9: 131430090-131500191 on Build GRCh38
Transcripts
NM_013318.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7385761 c.719G>T p.G240V Chr.9: 131446506-131446506 on Build GRCh38
4506863 c.731C>A p.S244* Chr.9: 131446518-131446518 on Build GRCh38
1245979 c.663G>A p.T221T Chr.9: 131446450-131446450 on Build GRCh38
4640898 c.683C>T p.T228I Chr.9: 131446470-131446470 on Build GRCh38
7103171 c.698G>C p.R233T Chr.9: 131446485-131446485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749745087 Chr.9: 131446385-131446385 on Build GRCh38
rs770952808 Chr.9: 131446418-131446418 on Build GRCh38
rs184006183 Chr.9: 131446450-131446450 on Build GRCh38
rs374961882 Chr.9: 131446459-131446459 on Build GRCh38
rs765201955 Chr.9: 131446345-131446345 on Build GRCh38
rs779271143 Chr.9: 131446503-131446503 on Build GRCh38
rs571964180 Chr.9: 131446430-131446430 on Build GRCh38
rs545470217 Chr.9: 131446436-131446436 on Build GRCh38
rs755452107 Chr.9: 131446498-131446498 on Build GRCh38
rs748187118 Chr.9: 131446390-131446390 on Build GRCh38
rs985704943 Chr.9: 131446481-131446481 on Build GRCh38
rs763847507 Chr.9: 131446469-131446469 on Build GRCh38
rs535303371 Chr.9: 131446382-131446382 on Build GRCh38
rs745581026 Chr.9: 131446363-131446363 on Build GRCh38
rs367606164 Chr.9: 131446488-131446488 on Build GRCh38
rs371842873 Chr.9: 131446447-131446447 on Build GRCh38
rs927325404 Chr.9: 131446494-131446494 on Build GRCh38
rs773300241 Chr.9: 131446406-131446406 on Build GRCh38
rs966803248 Chr.9: 131446377-131446378 on Build GRCh38
rs745328857 Chr.9: 131446368-131446368 on Build GRCh38
rs761020994 Chr.9: 131446428-131446428 on Build GRCh38
rs370331012 Chr.9: 131446360-131446360 on Build GRCh38
rs763942220 Chr.9: 131446449-131446449 on Build GRCh38
rs772249240 Chr.9: 131446403-131446403 on Build GRCh38
rs775165956 Chr.9: 131446350-131446350 on Build GRCh38
rs772017571 Chr.9: 131446518-131446518 on Build GRCh38
rs774417690 Chr.9: 131446387-131446387 on Build GRCh38
rs775450973 Chr.9: 131446431-131446431 on Build GRCh38
rs1055465643 Chr.9: 131446348-131446348 on Build GRCh38
rs373786036 Chr.9: 131446383-131446383 on Build GRCh38
rs778051277 Chr.9: 131446519-131446519 on Build GRCh38
rs756710623 Chr.9: 131446452-131446452 on Build GRCh38
rs750053072 Chr.9: 131446473-131446473 on Build GRCh38
rs1006688423 Chr.9: 131446349-131446349 on Build GRCh38
rs574422447 Chr.9: 131446357-131446357 on Build GRCh38
rs780356163 Chr.9: 131446491-131446491 on Build GRCh38
rs1003493611 Chr.9: 131446344-131446344 on Build GRCh38
rs762515504 Chr.9: 131446398-131446398 on Build GRCh38
rs774606986 Chr.9: 131446470-131446470 on Build GRCh38
rs368227837 Chr.9: 131446497-131446497 on Build GRCh38
rs531080097 Chr.9: 131446502-131446502 on Build GRCh38
rs371974492 Chr.9: 131446508-131446508 on Build GRCh38
rs76920830 Chr.9: 131446386-131446386 on Build GRCh38

View Full Product Details