Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54927596-54927825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCR1
Entrez Gene ID
9437
Gene Name
natural cytotoxicity triggering receptor 1
Gene Aliases
CD335, LY94, NK-p46, NKP46
UniGene
Hs.97084
Gene Chromosome Location
Chr.19: 54906042-54938211 on Build GRCh38
Transcripts
XM_011527530.2 NM_001127255.1 NM_139176.3 NM_206828.3 XM_006723075.3 XM_006723076.3 XM_011526596.2 XM_011526599.2 XM_011526601.2

Gene Symbol
NLRP7
Entrez Gene ID
199713
Gene Name
NLR family pyrin domain containing 7
Gene Aliases
CLR19.4, HYDM, NALP7, NOD12, PAN7, PYPAF3
UniGene
Hs.351118
Gene Chromosome Location
Chr.19: 54923509-54965184 on Build GRCh38
Transcripts
XM_011527530.2 NM_001127255.1 NM_139176.3 NM_206828.3 XM_006723075.3 XM_006723076.3 XM_011526596.2 XM_011526599.2 XM_011526601.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5671179 c.3063C>T p.L1021L Chr.19: 54927607-54927607 on Build GRCh38
1001292 c.2873G>T p.R958I Chr.19: 54927629-54927629 on Build GRCh38
5535486 c.2800G>A p.E934K Chr.19: 54927702-54927702 on Build GRCh38
1712833 c.2966C>T p.S989F Chr.19: 54927620-54927620 on Build GRCh38
6442555 c.2798T>C p.L933P Chr.19: 54927704-54927704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868850062 Chr.19: 54927671-54927671 on Build GRCh38
rs104895523 Chr.19: 54927695-54927695 on Build GRCh38
rs778669282 Chr.19: 54927723-54927723 on Build GRCh38
rs930057026 Chr.19: 54927740-54927740 on Build GRCh38
rs750756684 Chr.19: 54927687-54927687 on Build GRCh38
rs1045859520 Chr.19: 54927722-54927722 on Build GRCh38
rs269933 Chr.19: 54927798-54927798 on Build GRCh38
rs776999801 Chr.19: 54927771-54927771 on Build GRCh38
rs946073991 Chr.19: 54927678-54927678 on Build GRCh38
rs771221896 Chr.19: 54927751-54927751 on Build GRCh38
rs763415119 Chr.19: 54927641-54927641 on Build GRCh38
rs190983570 Chr.19: 54927775-54927775 on Build GRCh38
rs769969743 Chr.19: 54927777-54927777 on Build GRCh38
rs200945804 Chr.19: 54927790-54927790 on Build GRCh38
rs201545577 Chr.19: 54927730-54927730 on Build GRCh38
rs985978803 Chr.19: 54927659-54927659 on Build GRCh38
rs963309634 Chr.19: 54927713-54927713 on Build GRCh38
rs141473720 Chr.19: 54927747-54927747 on Build GRCh38
rs758308664 Chr.19: 54927737-54927737 on Build GRCh38
rs866843645 Chr.19: 54927670-54927670 on Build GRCh38
rs369045322 Chr.19: 54927792-54927792 on Build GRCh38
rs187895920 Chr.19: 54927668-54927668 on Build GRCh38
rs201472427 Chr.19: 54927802-54927802 on Build GRCh38
rs939239313 Chr.19: 54927754-54927754 on Build GRCh38
rs775870 Chr.19: 54927800-54927800 on Build GRCh38
rs766031898 Chr.19: 54927691-54927691 on Build GRCh38
rs202009680 Chr.19: 54927778-54927778 on Build GRCh38
rs777622728 Chr.19: 54927743-54927743 on Build GRCh38
rs774109007 Chr.19: 54927784-54927784 on Build GRCh38
rs776085217 Chr.19: 54927622-54927622 on Build GRCh38
rs1055434087 Chr.19: 54927761-54927761 on Build GRCh38
rs375946761 Chr.19: 54927721-54927721 on Build GRCh38
rs754674915 Chr.19: 54927805-54927805 on Build GRCh38
rs116109745 Chr.19: 54927803-54927803 on Build GRCh38
rs148164395 Chr.19: 54927649-54927649 on Build GRCh38
rs764485512 Chr.19: 54927642-54927642 on Build GRCh38
rs753277279 Chr.19: 54927718-54927718 on Build GRCh38
rs909155867 Chr.19: 54927686-54927686 on Build GRCh38

View Full Product Details