Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15479204-15479477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGLYRP2
Entrez Gene ID
114770
Gene Name
peptidoglycan recognition protein 2
Gene Aliases
HMFT0141, PGLYRPL, PGRP-L, PGRPL, TAGL-like, tagL, tagL-alpha, tagl-beta
UniGene
Hs.282244
Gene Chromosome Location
Chr.19: 15468645-15479504 on Build GRCh38
Transcripts
NM_052890.3 XM_006722633.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4956259 c.52C>T p.P18S Chr.19: 15479320-15479320 on Build GRCh38
4481341 c.24C>T p.I8I Chr.19: 15479348-15479348 on Build GRCh38
4956260 c.52C>T p.P18S Chr.19: 15479320-15479320 on Build GRCh38
1391154 c.25C>A p.L9I Chr.19: 15479347-15479347 on Build GRCh38
1391155 c.25C>A p.L9I Chr.19: 15479347-15479347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377710633 Chr.19: 15479258-15479258 on Build GRCh38
rs771520287 Chr.19: 15479388-15479389 on Build GRCh38
rs747537779 Chr.19: 15479310-15479315 on Build GRCh38
rs377661914 Chr.19: 15479406-15479406 on Build GRCh38
rs765539861 Chr.19: 15479395-15479395 on Build GRCh38
rs757090635 Chr.19: 15479325-15479325 on Build GRCh38
rs780454864 Chr.19: 15479440-15479440 on Build GRCh38
rs769267679 Chr.19: 15479371-15479371 on Build GRCh38
rs548711123 Chr.19: 15479259-15479259 on Build GRCh38
rs763233069 Chr.19: 15479254-15479254 on Build GRCh38
rs753902332 Chr.19: 15479447-15479447 on Build GRCh38
rs779749067 Chr.19: 15479421-15479422 on Build GRCh38
rs1016381996 Chr.19: 15479228-15479228 on Build GRCh38
rs759030891 Chr.19: 15479278-15479278 on Build GRCh38
rs1043814467 Chr.19: 15479380-15479380 on Build GRCh38
rs866766895 Chr.19: 15479451-15479452 on Build GRCh38
rs989624928 Chr.19: 15479310-15479310 on Build GRCh38
rs764791375 Chr.19: 15479279-15479279 on Build GRCh38
rs772542983 Chr.19: 15479361-15479361 on Build GRCh38
rs749400980 Chr.19: 15479351-15479351 on Build GRCh38
rs537669974 Chr.19: 15479386-15479386 on Build GRCh38
rs535072008 Chr.19: 15479256-15479256 on Build GRCh38
rs144264823 Chr.19: 15479245-15479245 on Build GRCh38
rs375566107 Chr.19: 15479287-15479287 on Build GRCh38
rs751574827 Chr.19: 15479415-15479415 on Build GRCh38
rs781259350 Chr.19: 15479341-15479341 on Build GRCh38
rs756232048 Chr.19: 15479344-15479344 on Build GRCh38
rs769244717 Chr.19: 15479356-15479356 on Build GRCh38
rs376070804 Chr.19: 15479429-15479429 on Build GRCh38
rs568666793 Chr.19: 15479363-15479363 on Build GRCh38
rs532388433 Chr.19: 15479241-15479241 on Build GRCh38
rs112159488 Chr.19: 15479365-15479365 on Build GRCh38
rs779477459 Chr.19: 15479357-15479357 on Build GRCh38
rs764053038 Chr.19: 15479298-15479298 on Build GRCh38
rs751249604 Chr.19: 15479318-15479318 on Build GRCh38
rs774945738 Chr.19: 15479372-15479372 on Build GRCh38
rs897807308 Chr.19: 15479233-15479233 on Build GRCh38
rs751645131 Chr.19: 15479428-15479428 on Build GRCh38
rs748401868 Chr.19: 15479358-15479358 on Build GRCh38
rs760344005 Chr.19: 15479303-15479303 on Build GRCh38
rs750725549 Chr.19: 15479342-15479342 on Build GRCh38
rs762832200 Chr.19: 15479292-15479292 on Build GRCh38
rs776356979 Chr.19: 15479274-15479274 on Build GRCh38
rs948579462 Chr.19: 15479234-15479234 on Build GRCh38
rs993535230 Chr.19: 15479235-15479235 on Build GRCh38
rs1030353916 Chr.19: 15479240-15479241 on Build GRCh38
rs767387133 Chr.19: 15479420-15479420 on Build GRCh38
rs114069253 Chr.19: 15479247-15479247 on Build GRCh38
rs112973534 Chr.19: 15479453-15479454 on Build GRCh38
rs557670900 Chr.19: 15479416-15479416 on Build GRCh38

View Full Product Details