Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 32207904-32208175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEP1B
Entrez Gene ID
4225
Gene Name
meprin A subunit beta
Gene Aliases
-
UniGene
Hs.194777
Gene Chromosome Location
Chr.18: 32190024-32222607 on Build GRCh38
Transcripts
NM_001308171.1 NM_005925.2 XM_011526013.2 XM_011526014.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1388426 c.798delT p.F267fs*9 Chr.18: 32208150-32208150 on Build GRCh38
4000394 c.792G>A p.S264S Chr.18: 32208144-32208144 on Build GRCh38
7401717 c.793T>G p.C265G Chr.18: 32208145-32208145 on Build GRCh38
7195591 c.785T>C p.M262T Chr.18: 32208137-32208137 on Build GRCh38
4071834 c.804A>G p.E268E Chr.18: 32208156-32208156 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759063440 Chr.18: 32208113-32208113 on Build GRCh38
rs777598097 Chr.18: 32208079-32208079 on Build GRCh38
rs147325009 Chr.18: 32208084-32208084 on Build GRCh38
rs370262667 Chr.18: 32208101-32208101 on Build GRCh38
rs776374473 Chr.18: 32208097-32208097 on Build GRCh38
rs768623995 Chr.18: 32207951-32207951 on Build GRCh38
rs960042165 Chr.18: 32207932-32207932 on Build GRCh38
rs910091880 Chr.18: 32207968-32207968 on Build GRCh38
rs531140945 Chr.18: 32207931-32207931 on Build GRCh38
rs917055697 Chr.18: 32207962-32207962 on Build GRCh38
rs984606317 Chr.18: 32207956-32207956 on Build GRCh38
rs368013600 Chr.18: 32208122-32208122 on Build GRCh38
rs373573919 Chr.18: 32208106-32208106 on Build GRCh38
rs35167501 Chr.18: 32208071-32208072 on Build GRCh38
rs186643040 Chr.18: 32207994-32207994 on Build GRCh38
rs973009642 Chr.18: 32208052-32208052 on Build GRCh38
rs4799640 Chr.18: 32208144-32208144 on Build GRCh38
rs991445674 Chr.18: 32207954-32207954 on Build GRCh38
rs567708209 Chr.18: 32207957-32207957 on Build GRCh38
rs772047490 Chr.18: 32208070-32208070 on Build GRCh38
rs1023474479 Chr.18: 32207950-32207950 on Build GRCh38
rs776045899 Chr.18: 32208125-32208125 on Build GRCh38
rs867548431 Chr.18: 32208015-32208015 on Build GRCh38
rs4799639 Chr.18: 32207937-32207937 on Build GRCh38

View Full Product Details