Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83858977-83859226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF27
Entrez Gene ID
55582
Gene Name
kinesin family member 27
Gene Aliases
-
UniGene
Hs.697514
Gene Chromosome Location
Chr.9: 83832795-83921539 on Build GRCh38
Transcripts
NM_001271927.1 NM_001271928.1 NM_017576.2 XM_011518848.2 XM_011518849.2 XM_011518850.2 XM_011518854.2 XM_011518856.2 XM_011518857.2 XM_011518858.2 XM_017014900.1 XM_017014901.1 XM_017014902.1 XM_017014903.1 XM_017014904.1 XM_017014905.1 XM_017014906.1 XM_017014907.1 XM_017014908.1 XM_017014909.1 XM_017014910.1 XM_017014911.1 XM_017014912.1 XM_017014913.1 XM_017014915.1 XM_017014916.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1674577 c.3085C>T p.Q1029* Chr.9: 83859221-83859221 on Build GRCh38
3375299 c.3120A>T p.K1040N Chr.9: 83859186-83859186 on Build GRCh38
6695403 c.3099C>T p.R1033R Chr.9: 83859207-83859207 on Build GRCh38
3764032 c.3106A>G p.N1036D Chr.9: 83859200-83859200 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776975627 Chr.9: 83859153-83859153 on Build GRCh38
rs768600707 Chr.9: 83859145-83859145 on Build GRCh38
rs755585990 Chr.9: 83859182-83859182 on Build GRCh38
rs140546026 Chr.9: 83859124-83859124 on Build GRCh38
rs369263386 Chr.9: 83859128-83859128 on Build GRCh38
rs191831133 Chr.9: 83859034-83859034 on Build GRCh38
rs775582206 Chr.9: 83859136-83859136 on Build GRCh38
rs761925902 Chr.9: 83859160-83859160 on Build GRCh38
rs753586591 Chr.9: 83859193-83859193 on Build GRCh38
rs751366959 Chr.9: 83859162-83859162 on Build GRCh38
rs372549064 Chr.9: 83859143-83859143 on Build GRCh38
rs112971484 Chr.9: 83859123-83859123 on Build GRCh38
rs752129956 Chr.9: 83859135-83859135 on Build GRCh38
rs530419565 Chr.9: 83859084-83859084 on Build GRCh38
rs534144528 Chr.9: 83859134-83859134 on Build GRCh38
rs371775428 Chr.9: 83859117-83859117 on Build GRCh38
rs375360469 Chr.9: 83859046-83859046 on Build GRCh38
rs766244397 Chr.9: 83859161-83859161 on Build GRCh38
rs147288435 Chr.9: 83859178-83859178 on Build GRCh38
rs746194165 Chr.9: 83859201-83859201 on Build GRCh38
rs777144395 Chr.9: 83859192-83859192 on Build GRCh38
rs759576962 Chr.9: 83859163-83859163 on Build GRCh38
rs149424110 Chr.9: 83859207-83859207 on Build GRCh38
rs55654273 Chr.9: 83859200-83859200 on Build GRCh38
rs201289514 Chr.9: 83859172-83859172 on Build GRCh38
rs980845597 Chr.9: 83859048-83859048 on Build GRCh38
rs746970797 Chr.9: 83859170-83859171 on Build GRCh38
rs568103213 Chr.9: 83859082-83859082 on Build GRCh38
rs988156628 Chr.9: 83859109-83859109 on Build GRCh38
rs548630870 Chr.9: 83859091-83859091 on Build GRCh38
rs756839697 Chr.9: 83859194-83859194 on Build GRCh38
rs767513341 Chr.9: 83859173-83859173 on Build GRCh38
rs777532252 Chr.9: 83859159-83859159 on Build GRCh38
rs757951697 Chr.9: 83859150-83859153 on Build GRCh38

View Full Product Details