Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 77344746-77345017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE8B
Entrez Gene ID
8622
Gene Name
phosphodiesterase 8B
Gene Aliases
ADSD, PPNAD3
UniGene
Hs.584830
Gene Chromosome Location
Chr.5: 77180252-77428256 on Build GRCh38
Transcripts
NM_001029851.2 NM_001029852.2 NM_001029853.2 NM_001029854.2 NM_003719.3 XM_005248621.4 XM_005248623.4 XM_005248624.4 XM_006714725.3 XM_006714726.3 XM_011543699.2 XM_011543700.2 XM_011543701.2 XM_011543702.2 XM_011543703.2 XM_011543704.2 XM_017010003.1 XM_017010004.1 XM_017010005.1 XM_017010006.1 XM_017010007.1 XM_017010008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3410418 c.855A>G p.T285T Chr.5: 77344910-77344910 on Build GRCh38
739201 c.874C>T p.Q292* Chr.5: 77344929-77344929 on Build GRCh38
6374252 c.818C>T p.T273I Chr.5: 77344873-77344873 on Build GRCh38
1666767 c.866A>G p.H289R Chr.5: 77344921-77344921 on Build GRCh38
295287 c.831C>G p.H277Q Chr.5: 77344886-77344886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370653898 Chr.5: 77344959-77344959 on Build GRCh38
rs774385824 Chr.5: 77344811-77344811 on Build GRCh38
rs752717678 Chr.5: 77344899-77344899 on Build GRCh38
rs767831002 Chr.5: 77344975-77344975 on Build GRCh38
rs777640717 Chr.5: 77344914-77344914 on Build GRCh38
rs1032706462 Chr.5: 77344787-77344787 on Build GRCh38
rs765306622 Chr.5: 77344842-77344842 on Build GRCh38
rs746578642 Chr.5: 77344923-77344923 on Build GRCh38
rs745428384 Chr.5: 77344941-77344941 on Build GRCh38
rs767492041 Chr.5: 77344829-77344829 on Build GRCh38
rs1015889040 Chr.5: 77344956-77344956 on Build GRCh38
rs781756640 Chr.5: 77344957-77344958 on Build GRCh38
rs753123305 Chr.5: 77344810-77344810 on Build GRCh38
rs201943184 Chr.5: 77344808-77344808 on Build GRCh38
rs748589757 Chr.5: 77344983-77344983 on Build GRCh38
rs575049091 Chr.5: 77344913-77344913 on Build GRCh38
rs756229238 Chr.5: 77344946-77344946 on Build GRCh38
rs377029423 Chr.5: 77344841-77344841 on Build GRCh38
rs768843534 Chr.5: 77344965-77344965 on Build GRCh38
rs368873268 Chr.5: 77344840-77344840 on Build GRCh38
rs1000013652 Chr.5: 77344786-77344786 on Build GRCh38
rs780551524 Chr.5: 77344935-77344935 on Build GRCh38
rs764549196 Chr.5: 77344810-77344810 on Build GRCh38
rs1037619020 Chr.5: 77344953-77344953 on Build GRCh38
rs368515727 Chr.5: 77344784-77344784 on Build GRCh38
rs369735697 Chr.5: 77344952-77344952 on Build GRCh38
rs762106858 Chr.5: 77344825-77344825 on Build GRCh38
rs537974977 Chr.5: 77344967-77344967 on Build GRCh38
rs144671246 Chr.5: 77344929-77344929 on Build GRCh38

View Full Product Details