Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 1019738-1019985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDI2
Entrez Gene ID
91734
Gene Name
isopentenyl-diphosphate delta isomerase 2
Gene Aliases
IPPI2
UniGene
Hs.591325
Gene Chromosome Location
Chr.10: 1018907-1025859 on Build GRCh38
Transcripts
NM_033261.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3396908 c.388T>A p.F130I Chr.10: 1019813-1019813 on Build GRCh38
3806408 c.425G>A p.R142K Chr.10: 1019776-1019776 on Build GRCh38
4691752 c.415A>C p.K139Q Chr.10: 1019786-1019786 on Build GRCh38
4832899 c.376G>C p.E126Q Chr.10: 1019825-1019825 on Build GRCh38
6530750 c.373C>G p.P125A Chr.10: 1019828-1019828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs10160004 Chr.10: 1019847-1019847 on Build GRCh38
rs113541732 Chr.10: 1019837-1019837 on Build GRCh38
rs773314383 Chr.10: 1019823-1019823 on Build GRCh38
rs767591457 Chr.10: 1019872-1019872 on Build GRCh38
rs1033557129 Chr.10: 1019950-1019950 on Build GRCh38
rs139607943 Chr.10: 1019802-1019802 on Build GRCh38
rs758543268 Chr.10: 1019800-1019801 on Build GRCh38
rs747567628 Chr.10: 1019865-1019888 on Build GRCh38
rs147373006 Chr.10: 1019782-1019782 on Build GRCh38
rs763333659 Chr.10: 1019853-1019853 on Build GRCh38
rs949410827 Chr.10: 1019784-1019784 on Build GRCh38
rs770349821 Chr.10: 1019951-1019951 on Build GRCh38
rs41294972 Chr.10: 1019764-1019764 on Build GRCh38
rs1016673041 Chr.10: 1019908-1019908 on Build GRCh38
rs771491159 Chr.10: 1019878-1019879 on Build GRCh38
rs777803253 Chr.10: 1019861-1019863 on Build GRCh38
rs576808585 Chr.10: 1019783-1019783 on Build GRCh38
rs774535404 Chr.10: 1019866-1019866 on Build GRCh38
rs764191760 Chr.10: 1019856-1019856 on Build GRCh38
rs766799448 Chr.10: 1019881-1019881 on Build GRCh38
rs41294974 Chr.10: 1019766-1019766 on Build GRCh38
rs746877846 Chr.10: 1019838-1019838 on Build GRCh38
rs572501482 Chr.10: 1019916-1019916 on Build GRCh38
rs752905367 Chr.10: 1019793-1019795 on Build GRCh38
rs1044261 Chr.10: 1019770-1019770 on Build GRCh38
rs771562485 Chr.10: 1019814-1019814 on Build GRCh38
rs905253524 Chr.10: 1019806-1019806 on Build GRCh38
rs372462801 Chr.10: 1019867-1019867 on Build GRCh38
rs780341800 Chr.10: 1019765-1019765 on Build GRCh38
rs898191140 Chr.10: 1019913-1019913 on Build GRCh38
rs1004395171 Chr.10: 1019883-1019883 on Build GRCh38
rs750931570 Chr.10: 1019875-1019875 on Build GRCh38
rs1002257816 Chr.10: 1019810-1019810 on Build GRCh38
rs368201188 Chr.10: 1019848-1019848 on Build GRCh38
rs756603336 Chr.10: 1019880-1019880 on Build GRCh38

View Full Product Details