Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 23566067-23566330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MKRN3
Entrez Gene ID
7681
Gene Name
makorin ring finger protein 3
Gene Aliases
CPPB2, D15S9, RNF63, ZFP127, ZNF127
UniGene
Hs.679587
Gene Chromosome Location
Chr.15: 23565307-23568020 on Build GRCh38
Transcripts
NM_005664.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
699502 c.295A>T p.I99F Chr.15: 23566077-23566077 on Build GRCh38
4999101 c.367G>T p.G123C Chr.15: 23566149-23566149 on Build GRCh38
699500 c.331G>T p.E111* Chr.15: 23566113-23566113 on Build GRCh38
699498 c.471G>C p.A157A Chr.15: 23566253-23566253 on Build GRCh38
959896 c.319G>A p.G107R Chr.15: 23566101-23566101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377146475 Chr.15: 23566265-23566265 on Build GRCh38
rs747691232 Chr.15: 23566277-23566277 on Build GRCh38
rs36072495 Chr.15: 23566175-23566175 on Build GRCh38
rs138394300 Chr.15: 23566098-23566098 on Build GRCh38
rs774020817 Chr.15: 23566197-23566197 on Build GRCh38
rs532500484 Chr.15: 23566185-23566185 on Build GRCh38
rs775857932 Chr.15: 23566261-23566261 on Build GRCh38
rs746805239 Chr.15: 23566101-23566101 on Build GRCh38
rs202210422 Chr.15: 23566154-23566154 on Build GRCh38
rs139225180 Chr.15: 23566297-23566297 on Build GRCh38
rs757781328 Chr.15: 23566278-23566278 on Build GRCh38
rs780368800 Chr.15: 23566293-23566293 on Build GRCh38
rs761098122 Chr.15: 23566143-23566143 on Build GRCh38
rs756140220 Chr.15: 23566272-23566272 on Build GRCh38
rs759878300 Chr.15: 23566208-23566208 on Build GRCh38
rs777747952 Chr.15: 23566100-23566100 on Build GRCh38
rs770089801 Chr.15: 23566260-23566260 on Build GRCh38
rs778487757 Chr.15: 23566276-23566276 on Build GRCh38
rs777034398 Chr.15: 23566222-23566222 on Build GRCh38
rs547482089 Chr.15: 23566192-23566192 on Build GRCh38
rs754708867 Chr.15: 23566241-23566241 on Build GRCh38
rs768221213 Chr.15: 23566196-23566196 on Build GRCh38
rs931206186 Chr.15: 23566190-23566190 on Build GRCh38
rs200032750 Chr.15: 23566153-23566153 on Build GRCh38
rs773676894 Chr.15: 23566242-23566243 on Build GRCh38
rs770320861 Chr.15: 23566284-23566284 on Build GRCh38
rs754655660 Chr.15: 23566275-23566275 on Build GRCh38
rs770695847 Chr.15: 23566115-23566115 on Build GRCh38
rs146108146 Chr.15: 23566263-23566263 on Build GRCh38
rs747489299 Chr.15: 23566258-23566258 on Build GRCh38
rs905116093 Chr.15: 23566181-23566181 on Build GRCh38
rs752762354 Chr.15: 23566165-23566165 on Build GRCh38
rs763195944 Chr.15: 23566257-23566258 on Build GRCh38
rs369442499 Chr.15: 23566304-23566304 on Build GRCh38
rs762104625 Chr.15: 23566253-23566253 on Build GRCh38
rs536586920 Chr.15: 23566210-23566210 on Build GRCh38
rs886965228 Chr.15: 23566169-23566169 on Build GRCh38
rs199803521 Chr.15: 23566243-23566243 on Build GRCh38
rs768856943 Chr.15: 23566299-23566299 on Build GRCh38
rs773786543 Chr.15: 23566142-23566142 on Build GRCh38
rs780006913 Chr.15: 23566237-23566237 on Build GRCh38
rs372884408 Chr.15: 23566306-23566306 on Build GRCh38
rs778533814 Chr.15: 23566242-23566242 on Build GRCh38
rs757168649 Chr.15: 23566177-23566177 on Build GRCh38
rs746391893 Chr.15: 23566259-23566259 on Build GRCh38
rs764114571 Chr.15: 23566217-23566217 on Build GRCh38
rs762441841 Chr.15: 23566222-23566222 on Build GRCh38
rs146391768 Chr.15: 23566128-23566128 on Build GRCh38
rs375745454 Chr.15: 23566310-23566310 on Build GRCh38
rs200402711 Chr.15: 23566264-23566264 on Build GRCh38
rs552696758 Chr.15: 23566274-23566274 on Build GRCh38
rs765492026 Chr.15: 23566160-23566160 on Build GRCh38
rs144864886 Chr.15: 23566207-23566207 on Build GRCh38
rs536725835 Chr.15: 23566238-23566238 on Build GRCh38
rs781005844 Chr.15: 23566180-23566180 on Build GRCh38
rs370156382 Chr.15: 23566191-23566191 on Build GRCh38
rs775113089 Chr.15: 23566136-23566136 on Build GRCh38
rs757192830 Chr.15: 23566223-23566223 on Build GRCh38
rs202089304 Chr.15: 23566152-23566152 on Build GRCh38
rs777359232 Chr.15: 23566282-23566282 on Build GRCh38
rs771456272 Chr.15: 23566247-23566247 on Build GRCh38
rs772698198 Chr.15: 23566140-23566140 on Build GRCh38
rs148583347 Chr.15: 23566225-23566225 on Build GRCh38
rs142966356 Chr.15: 23566226-23566226 on Build GRCh38
rs900932835 Chr.15: 23566148-23566148 on Build GRCh38
rs372340447 Chr.15: 23566262-23566262 on Build GRCh38
rs774367502 Chr.15: 23566301-23566301 on Build GRCh38
rs1039350043 Chr.15: 23566144-23566144 on Build GRCh38
rs745694373 Chr.15: 23566183-23566183 on Build GRCh38
rs368059154 Chr.15: 23566213-23566213 on Build GRCh38
rs143897856 Chr.15: 23566099-23566099 on Build GRCh38
rs149344478 Chr.15: 23566184-23566184 on Build GRCh38
rs776217596 Chr.15: 23566211-23566211 on Build GRCh38
rs199845546 Chr.15: 23566139-23566139 on Build GRCh38
rs1033966658 Chr.15: 23566132-23566132 on Build GRCh38
rs747509730 Chr.15: 23566205-23566205 on Build GRCh38
rs569949058 Chr.15: 23566229-23566229 on Build GRCh38
rs765790834 Chr.15: 23566224-23566225 on Build GRCh38
rs745479517 Chr.15: 23566129-23566129 on Build GRCh38
rs1004369260 Chr.15: 23566215-23566215 on Build GRCh38
rs750269411 Chr.15: 23566269-23566269 on Build GRCh38
rs371642799 Chr.15: 23566216-23566216 on Build GRCh38
rs1011130063 Chr.15: 23566286-23566286 on Build GRCh38
rs201431738 Chr.15: 23566176-23566176 on Build GRCh38
rs766235976 Chr.15: 23566273-23566273 on Build GRCh38
rs777074455 Chr.15: 23566257-23566257 on Build GRCh38
rs926211093 Chr.15: 23566133-23566133 on Build GRCh38
rs1005371940 Chr.15: 23566173-23566173 on Build GRCh38

View Full Product Details