Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124542876-124543147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8B12
Entrez Gene ID
219858
Gene Name
olfactory receptor family 8 subfamily B member 12
Gene Aliases
OR11-317
UniGene
Hs.553634
Gene Chromosome Location
Chr.11: 124542722-124543654 on Build GRCh38
Transcripts
NM_001005195.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
313774 c.676C>A p.H226N Chr.11: 124542979-124542979 on Build GRCh38
5600126 c.544C>T p.P182S Chr.11: 124543111-124543111 on Build GRCh38
5595053 c.639C>T p.V213V Chr.11: 124543016-124543016 on Build GRCh38
5613315 c.679A>T p.N227Y Chr.11: 124542976-124542976 on Build GRCh38
5676731 c.667A>T p.S223C Chr.11: 124542988-124542988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984938943 Chr.11: 124542992-124542992 on Build GRCh38
rs766806750 Chr.11: 124542949-124542949 on Build GRCh38
rs199807361 Chr.11: 124542925-124542925 on Build GRCh38
rs774922867 Chr.11: 124543077-124543077 on Build GRCh38
rs745810160 Chr.11: 124543073-124543073 on Build GRCh38
rs755600196 Chr.11: 124542956-124542956 on Build GRCh38
rs1045624126 Chr.11: 124543030-124543030 on Build GRCh38
rs372694614 Chr.11: 124543070-124543070 on Build GRCh38
rs766638642 Chr.11: 124543074-124543074 on Build GRCh38
rs923545085 Chr.11: 124543090-124543090 on Build GRCh38
rs374139726 Chr.11: 124542916-124542916 on Build GRCh38
rs376653327 Chr.11: 124543071-124543071 on Build GRCh38
rs61747005 Chr.11: 124542942-124542942 on Build GRCh38
rs756968192 Chr.11: 124543018-124543018 on Build GRCh38
rs139856739 Chr.11: 124543062-124543062 on Build GRCh38
rs760051788 Chr.11: 124543086-124543086 on Build GRCh38
rs776638132 Chr.11: 124543040-124543040 on Build GRCh38
rs752257928 Chr.11: 124542969-124542969 on Build GRCh38
rs748250380 Chr.11: 124543043-124543043 on Build GRCh38
rs777311594 Chr.11: 124542988-124542988 on Build GRCh38
rs939411629 Chr.11: 124543054-124543054 on Build GRCh38
rs768151345 Chr.11: 124543089-124543089 on Build GRCh38
rs745805931 Chr.11: 124543038-124543038 on Build GRCh38
rs757069212 Chr.11: 124542941-124542941 on Build GRCh38
rs755768568 Chr.11: 124542975-124542975 on Build GRCh38
rs34165015 Chr.11: 124542963-124542964 on Build GRCh38
rs752202804 Chr.11: 124543093-124543093 on Build GRCh38
rs368885076 Chr.11: 124542936-124542936 on Build GRCh38
rs774183192 Chr.11: 124542920-124542920 on Build GRCh38
rs759506849 Chr.11: 124542926-124542926 on Build GRCh38
rs770019515 Chr.11: 124543051-124543051 on Build GRCh38
rs147488884 Chr.11: 124543039-124543039 on Build GRCh38
rs768713495 Chr.11: 124542930-124542930 on Build GRCh38
rs549372512 Chr.11: 124542948-124542948 on Build GRCh38
rs767131052 Chr.11: 124542967-124542967 on Build GRCh38
rs778829590 Chr.11: 124543020-124543020 on Build GRCh38
rs948216136 Chr.11: 124543029-124543029 on Build GRCh38
rs143059318 Chr.11: 124542955-124542955 on Build GRCh38
rs749067224 Chr.11: 124543001-124543001 on Build GRCh38
rs915401329 Chr.11: 124542993-124542993 on Build GRCh38

View Full Product Details