Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 133150857-133151090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDH10
Entrez Gene ID
57575
Gene Name
protocadherin 10
Gene Aliases
OL-PCDH, PCDH19
UniGene
Hs.192859
Gene Chromosome Location
Chr.4: 133149290-133208606 on Build GRCh38
Transcripts
NM_020815.2 NM_032961.2 XM_011532150.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
585830 c.750C>T p.F250F Chr.4: 133150890-133150890 on Build GRCh38
6323249 c.829C>A p.P277T Chr.4: 133150969-133150969 on Build GRCh38
1427087 c.881C>T p.S294L Chr.4: 133151021-133151021 on Build GRCh38
1051220 c.734A>G p.D245G Chr.4: 133150874-133150874 on Build GRCh38
732595 c.900T>C p.L300L Chr.4: 133151040-133151040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369263513 Chr.4: 133150977-133150977 on Build GRCh38
rs375479426 Chr.4: 133150893-133150893 on Build GRCh38
rs772098781 Chr.4: 133150992-133150992 on Build GRCh38
rs756420365 Chr.4: 133150985-133150985 on Build GRCh38
rs371314024 Chr.4: 133151043-133151043 on Build GRCh38
rs376487876 Chr.4: 133150937-133150937 on Build GRCh38
rs762172525 Chr.4: 133150941-133150941 on Build GRCh38
rs767573925 Chr.4: 133151044-133151044 on Build GRCh38
rs763406461 Chr.4: 133151023-133151023 on Build GRCh38
rs781420676 Chr.4: 133150884-133150884 on Build GRCh38
rs760135106 Chr.4: 133150940-133150940 on Build GRCh38
rs376916059 Chr.4: 133151025-133151025 on Build GRCh38
rs746462897 Chr.4: 133150995-133150995 on Build GRCh38
rs139977349 Chr.4: 133150932-133150932 on Build GRCh38
rs765513404 Chr.4: 133150944-133150944 on Build GRCh38
rs372547042 Chr.4: 133150986-133150986 on Build GRCh38
rs986553655 Chr.4: 133150891-133150891 on Build GRCh38
rs760293091 Chr.4: 133150933-133150933 on Build GRCh38
rs370947882 Chr.4: 133151068-133151068 on Build GRCh38
rs575638492 Chr.4: 133150935-133150935 on Build GRCh38
rs546430185 Chr.4: 133151038-133151038 on Build GRCh38
rs199927426 Chr.4: 133150879-133150879 on Build GRCh38
rs761355771 Chr.4: 133151000-133151000 on Build GRCh38
rs746941061 Chr.4: 133151065-133151065 on Build GRCh38
rs752872197 Chr.4: 133151050-133151050 on Build GRCh38
rs780072459 Chr.4: 133151064-133151064 on Build GRCh38
rs936994371 Chr.4: 133151070-133151070 on Build GRCh38
rs758809294 Chr.4: 133150953-133150953 on Build GRCh38
rs566769897 Chr.4: 133151060-133151060 on Build GRCh38
rs375893532 Chr.4: 133151057-133151057 on Build GRCh38
rs756259780 Chr.4: 133151051-133151051 on Build GRCh38
rs751594784 Chr.4: 133150969-133150969 on Build GRCh38
rs773414657 Chr.4: 133151014-133151014 on Build GRCh38
rs766911892 Chr.4: 133150959-133150959 on Build GRCh38
rs745716961 Chr.4: 133150987-133150987 on Build GRCh38
rs538495127 Chr.4: 133151016-133151016 on Build GRCh38
rs748316231 Chr.4: 133150981-133150981 on Build GRCh38
rs774272199 Chr.4: 133150920-133150920 on Build GRCh38
rs144002604 Chr.4: 133150878-133150878 on Build GRCh38
rs754982520 Chr.4: 133151034-133151034 on Build GRCh38
rs149439832 Chr.4: 133150974-133150974 on Build GRCh38
rs778087588 Chr.4: 133151056-133151056 on Build GRCh38
rs752073353 Chr.4: 133151029-133151029 on Build GRCh38
rs764985616 Chr.4: 133151009-133151009 on Build GRCh38
rs147313392 Chr.4: 133150931-133150931 on Build GRCh38
rs370972266 Chr.4: 133151007-133151007 on Build GRCh38
rs775594011 Chr.4: 133150993-133150993 on Build GRCh38
rs754949546 Chr.4: 133150996-133150996 on Build GRCh38
rs562154592 Chr.4: 133150899-133150899 on Build GRCh38

View Full Product Details