Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47625567-47625828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTCH2
Entrez Gene ID
23788
Gene Name
mitochondrial carrier 2
Gene Aliases
HSPC032, MIMP, SLC25A50
UniGene
Hs.269944
Gene Chromosome Location
Chr.11: 47604309-47642654 on Build GRCh38
Transcripts
NM_001317231.1 NM_001317232.1 NM_001317233.1 NM_014342.3 XM_006718172.2 XM_011519959.2 XM_011519960.2 XM_011519961.2 XM_017017462.1 XM_017017463.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
927467 c.743A>T p.N248I Chr.11: 47625680-47625680 on Build GRCh38
5147514 c.692G>A p.S231N Chr.11: 47625731-47625731 on Build GRCh38
6461822 c.690G>C p.A230A Chr.11: 47625733-47625733 on Build GRCh38
5021716 c.737T>C p.V246A Chr.11: 47625686-47625686 on Build GRCh38
5372558 c.744C>T p.N248N Chr.11: 47625679-47625679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913406091 Chr.11: 47625733-47625733 on Build GRCh38
rs376983287 Chr.11: 47625624-47625624 on Build GRCh38
rs761207064 Chr.11: 47625784-47625784 on Build GRCh38
rs946266542 Chr.11: 47625750-47625750 on Build GRCh38
rs756016633 Chr.11: 47625660-47625661 on Build GRCh38
rs936092642 Chr.11: 47625695-47625695 on Build GRCh38
rs184871712 Chr.11: 47625648-47625648 on Build GRCh38
rs76666113 Chr.11: 47625713-47625713 on Build GRCh38
rs753559463 Chr.11: 47625626-47625626 on Build GRCh38
rs771587009 Chr.11: 47625661-47625661 on Build GRCh38
rs188681664 Chr.11: 47625769-47625769 on Build GRCh38
rs980104644 Chr.11: 47625632-47625632 on Build GRCh38
rs753583175 Chr.11: 47625790-47625810 on Build GRCh38
rs267602908 Chr.11: 47625717-47625717 on Build GRCh38
rs753261769 Chr.11: 47625783-47625783 on Build GRCh38
rs746941936 Chr.11: 47625671-47625671 on Build GRCh38
rs267602907 Chr.11: 47625716-47625716 on Build GRCh38
rs1041011193 Chr.11: 47625782-47625782 on Build GRCh38
rs544100641 Chr.11: 47625698-47625698 on Build GRCh38
rs771102156 Chr.11: 47625732-47625732 on Build GRCh38
rs768441220 Chr.11: 47625691-47625691 on Build GRCh38
rs750426344 Chr.11: 47625642-47625642 on Build GRCh38
rs911325180 Chr.11: 47625751-47625751 on Build GRCh38
rs756811078 Chr.11: 47625627-47625627 on Build GRCh38
rs774613234 Chr.11: 47625734-47625734 on Build GRCh38
rs767632456 Chr.11: 47625780-47625780 on Build GRCh38
rs773071465 Chr.11: 47625619-47625619 on Build GRCh38
rs78071782 Chr.11: 47625686-47625686 on Build GRCh38
rs894789875 Chr.11: 47625611-47625611 on Build GRCh38
rs764575237 Chr.11: 47625788-47625788 on Build GRCh38
rs779911649 Chr.11: 47625663-47625663 on Build GRCh38
rs779859007 Chr.11: 47625784-47625788 on Build GRCh38
rs961672783 Chr.11: 47625643-47625643 on Build GRCh38
rs376518855 Chr.11: 47625770-47625770 on Build GRCh38

View Full Product Details