Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57029569-57029824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO1A
Entrez Gene ID
4640
Gene Name
myosin IA
Gene Aliases
BBMI, DFNA48, MIHC, MYHL
UniGene
Hs.5394
Gene Chromosome Location
Chr.12: 57028517-57050765 on Build GRCh38
Transcripts
NM_001256041.1 NM_005379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1363204 c.2710C>T p.R904C Chr.12: 57029754-57029754 on Build GRCh38
5051804 c.2695G>A p.V899M Chr.12: 57029769-57029769 on Build GRCh38
6137399 c.2671_2672GG>CT p.G891L Chr.12: 57029792-57029793 on Build GRCh38
6442400 c.2665G>A p.E889K Chr.12: 57029799-57029799 on Build GRCh38
6435499 c.2704G>T p.V902F Chr.12: 57029760-57029760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148582008 Chr.12: 57029780-57029780 on Build GRCh38
rs59514028 Chr.12: 57029712-57029712 on Build GRCh38
rs146373415 Chr.12: 57029756-57029756 on Build GRCh38
rs746997377 Chr.12: 57029608-57029608 on Build GRCh38
rs75239044 Chr.12: 57029623-57029623 on Build GRCh38
rs536778643 Chr.12: 57029784-57029784 on Build GRCh38
rs373891069 Chr.12: 57029711-57029711 on Build GRCh38
rs769911842 Chr.12: 57029773-57029773 on Build GRCh38
rs758088084 Chr.12: 57029754-57029754 on Build GRCh38
rs759878403 Chr.12: 57029799-57029799 on Build GRCh38
rs375656169 Chr.12: 57029595-57029595 on Build GRCh38
rs776703736 Chr.12: 57029629-57029629 on Build GRCh38
rs76849707 Chr.12: 57029622-57029622 on Build GRCh38
rs199692652 Chr.12: 57029766-57029766 on Build GRCh38
rs200448145 Chr.12: 57029593-57029593 on Build GRCh38
rs772061719 Chr.12: 57029596-57029596 on Build GRCh38
rs55985817 Chr.12: 57029733-57029733 on Build GRCh38
rs776667897 Chr.12: 57029755-57029755 on Build GRCh38
rs368656151 Chr.12: 57029721-57029721 on Build GRCh38
rs79722452 Chr.12: 57029620-57029620 on Build GRCh38
rs751910306 Chr.12: 57029713-57029715 on Build GRCh38
rs768104446 Chr.12: 57029803-57029803 on Build GRCh38
rs200861737 Chr.12: 57029741-57029741 on Build GRCh38
rs370491058 Chr.12: 57029597-57029597 on Build GRCh38
rs747166970 Chr.12: 57029647-57029647 on Build GRCh38
rs751974348 Chr.12: 57029704-57029704 on Build GRCh38
rs886049695 Chr.12: 57029746-57029746 on Build GRCh38
rs755412826 Chr.12: 57029761-57029763 on Build GRCh38
rs1010769720 Chr.12: 57029804-57029804 on Build GRCh38
rs79375467 Chr.12: 57029619-57029619 on Build GRCh38
rs376674552 Chr.12: 57029769-57029769 on Build GRCh38
rs529506371 Chr.12: 57029637-57029637 on Build GRCh38
rs750119242 Chr.12: 57029753-57029753 on Build GRCh38
rs1053322144 Chr.12: 57029685-57029685 on Build GRCh38
rs773374837 Chr.12: 57029777-57029777 on Build GRCh38
rs928512499 Chr.12: 57029641-57029641 on Build GRCh38
rs766792590 Chr.12: 57029692-57029692 on Build GRCh38
rs778682754 Chr.12: 57029744-57029744 on Build GRCh38
rs543945564 Chr.12: 57029677-57029677 on Build GRCh38
rs777541281 Chr.12: 57029790-57029791 on Build GRCh38
rs770001309 Chr.12: 57029634-57029634 on Build GRCh38
rs774795013 Chr.12: 57029796-57029796 on Build GRCh38
rs933169836 Chr.12: 57029720-57029720 on Build GRCh38
rs748197044 Chr.12: 57029770-57029770 on Build GRCh38

View Full Product Details