Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 11689549-11689780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAL
Entrez Gene ID
2774
Gene Name
G protein subunit alpha L
Gene Aliases
DYT25
UniGene
Hs.136295
Gene Chromosome Location
Chr.18: 11688956-11885685 on Build GRCh38
Transcripts
NM_182978.3 XM_006722324.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5696995 c.53A>C p.D18A Chr.18: 11689616-11689616 on Build GRCh38
6556849 c.102G>T p.P34P Chr.18: 11689665-11689665 on Build GRCh38
5959648 c.107_108insGGCCCT p.A39_P40insLA Chr.18: 11689670-11689671 on Build GRCh38
4130911 c.107C>T p.P36L Chr.18: 11689670-11689670 on Build GRCh38
3749432 c.137G>A p.R46K Chr.18: 11689700-11689700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017206708 Chr.18: 11689652-11689652 on Build GRCh38
rs1023317715 Chr.18: 11689604-11689604 on Build GRCh38
rs564907176 Chr.18: 11689713-11689713 on Build GRCh38
rs964964441 Chr.18: 11689656-11689656 on Build GRCh38
rs954433506 Chr.18: 11689623-11689623 on Build GRCh38
rs779349517 Chr.18: 11689761-11689761 on Build GRCh38
rs1032625784 Chr.18: 11689600-11689600 on Build GRCh38
rs1044393018 Chr.18: 11689568-11689568 on Build GRCh38
rs901005939 Chr.18: 11689740-11689740 on Build GRCh38
rs531745431 Chr.18: 11689676-11689677 on Build GRCh38
rs987117042 Chr.18: 11689632-11689632 on Build GRCh38
rs893639863 Chr.18: 11689601-11689601 on Build GRCh38
rs750015298 Chr.18: 11689695-11689695 on Build GRCh38
rs755250899 Chr.18: 11689745-11689745 on Build GRCh38
rs201898548 Chr.18: 11689670-11689671 on Build GRCh38
rs7236061 Chr.18: 11689670-11689670 on Build GRCh38
rs543723400 Chr.18: 11689705-11689705 on Build GRCh38
rs767065649 Chr.18: 11689683-11689683 on Build GRCh38
rs753562019 Chr.18: 11689724-11689724 on Build GRCh38
rs956895925 Chr.18: 11689626-11689626 on Build GRCh38
rs532055871 Chr.18: 11689701-11689701 on Build GRCh38
rs768781193 Chr.18: 11689684-11689684 on Build GRCh38
rs905132944 Chr.18: 11689572-11689572 on Build GRCh38
rs7236433 Chr.18: 11689700-11689700 on Build GRCh38
rs1002109765 Chr.18: 11689575-11689575 on Build GRCh38
rs1005314490 Chr.18: 11689757-11689757 on Build GRCh38
rs982367401 Chr.18: 11689617-11689617 on Build GRCh38
rs577163323 Chr.18: 11689671-11689676 on Build GRCh38
rs1005653996 Chr.18: 11689578-11689578 on Build GRCh38
rs993990807 Chr.18: 11689749-11689749 on Build GRCh38
rs1040980055 Chr.18: 11689726-11689726 on Build GRCh38
rs543230597 Chr.18: 11689611-11689611 on Build GRCh38
rs915337719 Chr.18: 11689664-11689664 on Build GRCh38
rs937952329 Chr.18: 11689737-11689737 on Build GRCh38
rs952400171 Chr.18: 11689576-11689576 on Build GRCh38
rs917356429 Chr.18: 11689706-11689706 on Build GRCh38
rs950092521 Chr.18: 11689711-11689711 on Build GRCh38
rs1009777479 Chr.18: 11689609-11689609 on Build GRCh38
rs1044443716 Chr.18: 11689719-11689719 on Build GRCh38
rs1046793623 Chr.18: 11689756-11689756 on Build GRCh38
rs920804434 Chr.18: 11689688-11689688 on Build GRCh38
rs761851127 Chr.18: 11689680-11689680 on Build GRCh38
rs910562901 Chr.18: 11689715-11689715 on Build GRCh38
rs767243364 Chr.18: 11689659-11689660 on Build GRCh38

View Full Product Details