Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70532462-70532723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PALD1
Entrez Gene ID
27143
Gene Name
phosphatase domain containing, paladin 1
Gene Aliases
KIAA1274, PALD
UniGene
Hs.202351
Gene Chromosome Location
Chr.10: 70458542-70568450 on Build GRCh38
Transcripts
NM_014431.2 XM_011539637.2 XM_011539638.2 XM_017016072.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
920013 c.634-1G>A p.? Chr.10: 70532620-70532620 on Build GRCh38
1348838 c.718G>A p.A240T Chr.10: 70532705-70532705 on Build GRCh38
6066530 c.648C>A p.A216A Chr.10: 70532635-70532635 on Build GRCh38
7155698 c.723C>G p.I241M Chr.10: 70532710-70532710 on Build GRCh38
6482680 c.720C>G p.A240A Chr.10: 70532707-70532707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759381672 Chr.10: 70532647-70532647 on Build GRCh38
rs539993222 Chr.10: 70532593-70532593 on Build GRCh38
rs10999378 Chr.10: 70532502-70532502 on Build GRCh38
rs747742327 Chr.10: 70532623-70532623 on Build GRCh38
rs780554144 Chr.10: 70532603-70532603 on Build GRCh38
rs535061702 Chr.10: 70532495-70532495 on Build GRCh38
rs774272391 Chr.10: 70532645-70532645 on Build GRCh38
rs538988042 Chr.10: 70532545-70532545 on Build GRCh38
rs769447017 Chr.10: 70532686-70532686 on Build GRCh38
rs1049737966 Chr.10: 70532536-70532536 on Build GRCh38
rs376346914 Chr.10: 70532674-70532674 on Build GRCh38
rs370081067 Chr.10: 70532620-70532620 on Build GRCh38
rs946279625 Chr.10: 70532523-70532523 on Build GRCh38
rs769539071 Chr.10: 70532626-70532626 on Build GRCh38
rs572000263 Chr.10: 70532503-70532503 on Build GRCh38
rs750659533 Chr.10: 70532668-70532668 on Build GRCh38
rs145337663 Chr.10: 70532685-70532685 on Build GRCh38
rs191087909 Chr.10: 70532693-70532693 on Build GRCh38
rs186538437 Chr.10: 70532490-70532490 on Build GRCh38
rs767544469 Chr.10: 70532651-70532651 on Build GRCh38
rs765395228 Chr.10: 70532667-70532667 on Build GRCh38
rs573290904 Chr.10: 70532652-70532652 on Build GRCh38
rs773025697 Chr.10: 70532634-70532634 on Build GRCh38
rs993050380 Chr.10: 70532485-70532485 on Build GRCh38
rs74139680 Chr.10: 70532611-70532611 on Build GRCh38
rs748950169 Chr.10: 70532701-70532701 on Build GRCh38
rs146140803 Chr.10: 70532644-70532644 on Build GRCh38
rs944165236 Chr.10: 70532566-70532566 on Build GRCh38
rs936157015 Chr.10: 70532559-70532559 on Build GRCh38
rs780275802 Chr.10: 70532675-70532675 on Build GRCh38
rs557493069 Chr.10: 70532556-70532556 on Build GRCh38
rs568396036 Chr.10: 70532573-70532573 on Build GRCh38
rs373865797 Chr.10: 70532658-70532658 on Build GRCh38
rs987125995 Chr.10: 70532500-70532500 on Build GRCh38
rs925882614 Chr.10: 70532511-70532511 on Build GRCh38
rs747462790 Chr.10: 70532676-70532676 on Build GRCh38
rs757360581 Chr.10: 70532664-70532664 on Build GRCh38
rs1041948758 Chr.10: 70532554-70532554 on Build GRCh38
rs1055827764 Chr.10: 70532596-70532596 on Build GRCh38
rs142191615 Chr.10: 70532689-70532689 on Build GRCh38
rs747476216 Chr.10: 70532608-70532608 on Build GRCh38

View Full Product Details