Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34300886-34301131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0355
Entrez Gene ID
9710
Gene Name
KIAA0355
Gene Aliases
-
UniGene
Hs.330073
Gene Chromosome Location
Chr.19: 34254551-34355566 on Build GRCh38
Transcripts
NM_014686.3 XM_005259444.1 XM_017027525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5964878 c.495+2T>C p.? Chr.19: 34300970-34300970 on Build GRCh38
234256 c.466C>T p.Q156* Chr.19: 34300939-34300939 on Build GRCh38
3532290 c.472G>A p.E158K Chr.19: 34300945-34300945 on Build GRCh38
4493574 c.417C>T p.I139I Chr.19: 34300890-34300890 on Build GRCh38
6084580 c.426G>T p.M142I Chr.19: 34300899-34300899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768101617 Chr.19: 34300977-34300977 on Build GRCh38
rs62122214 Chr.19: 34300917-34300917 on Build GRCh38
rs891316055 Chr.19: 34301057-34301057 on Build GRCh38
rs773154057 Chr.19: 34301006-34301006 on Build GRCh38
rs893745805 Chr.19: 34301095-34301095 on Build GRCh38
rs764603418 Chr.19: 34300935-34300935 on Build GRCh38
rs754107575 Chr.19: 34300913-34300913 on Build GRCh38
rs932936766 Chr.19: 34301007-34301007 on Build GRCh38
rs529496099 Chr.19: 34300973-34300973 on Build GRCh38
rs1052346063 Chr.19: 34301087-34301087 on Build GRCh38
rs376530523 Chr.19: 34300990-34300990 on Build GRCh38
rs950097548 Chr.19: 34300997-34300997 on Build GRCh38
rs779391650 Chr.19: 34300972-34300972 on Build GRCh38
rs746152945 Chr.19: 34300978-34300978 on Build GRCh38
rs370579561 Chr.19: 34301017-34301017 on Build GRCh38
rs146643118 Chr.19: 34300934-34300934 on Build GRCh38
rs566381372 Chr.19: 34301062-34301062 on Build GRCh38
rs757616651 Chr.19: 34300947-34300947 on Build GRCh38
rs755800148 Chr.19: 34300963-34300963 on Build GRCh38
rs752106417 Chr.19: 34300940-34300940 on Build GRCh38
rs551356719 Chr.19: 34301077-34301077 on Build GRCh38
rs528640076 Chr.19: 34301107-34301107 on Build GRCh38
rs181213766 Chr.19: 34301094-34301094 on Build GRCh38
rs752822969 Chr.19: 34300927-34300927 on Build GRCh38
rs759859175 Chr.19: 34300916-34300916 on Build GRCh38
rs759610631 Chr.19: 34301009-34301009 on Build GRCh38
rs765523205 Chr.19: 34301010-34301010 on Build GRCh38
rs781598440 Chr.19: 34300949-34300949 on Build GRCh38
rs763063958 Chr.19: 34301016-34301016 on Build GRCh38
rs571345352 Chr.19: 34301089-34301089 on Build GRCh38
rs745469726 Chr.19: 34300951-34300951 on Build GRCh38
rs774401718 Chr.19: 34300985-34300985 on Build GRCh38
rs146007478 Chr.19: 34301066-34301066 on Build GRCh38
rs775380576 Chr.19: 34301011-34301011 on Build GRCh38

View Full Product Details