Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42234482-42234711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSK3A
Entrez Gene ID
2931
Gene Name
glycogen synthase kinase 3 alpha
Gene Aliases
-
UniGene
Hs.466828
Gene Chromosome Location
Chr.19: 42230186-42243330 on Build GRCh38
Transcripts
NM_019884.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6383567 c.776T>C p.L259P Chr.19: 42234569-42234569 on Build GRCh38
4154227 c.688C>G p.R230G Chr.19: 42234657-42234657 on Build GRCh38
48412 c.688C>T p.R230C Chr.19: 42234657-42234657 on Build GRCh38
712696 c.784T>C p.C262R Chr.19: 42234561-42234561 on Build GRCh38
439638 c.710C>T p.S237F Chr.19: 42234635-42234635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775097746 Chr.19: 42234535-42234535 on Build GRCh38
rs986885157 Chr.19: 42234677-42234677 on Build GRCh38
rs770210270 Chr.19: 42234503-42234503 on Build GRCh38
rs759265893 Chr.19: 42234671-42234671 on Build GRCh38
rs764143628 Chr.19: 42234559-42234559 on Build GRCh38
rs575634045 Chr.19: 42234686-42234686 on Build GRCh38
rs745584114 Chr.19: 42234510-42234510 on Build GRCh38
rs908624912 Chr.19: 42234641-42234641 on Build GRCh38
rs760058249 Chr.19: 42234537-42234537 on Build GRCh38
rs778151740 Chr.19: 42234682-42234682 on Build GRCh38
rs879265393 Chr.19: 42234527-42234527 on Build GRCh38
rs756593040 Chr.19: 42234676-42234676 on Build GRCh38
rs373773536 Chr.19: 42234586-42234586 on Build GRCh38
rs750207489 Chr.19: 42234588-42234588 on Build GRCh38
rs751671051 Chr.19: 42234628-42234628 on Build GRCh38
rs781244052 Chr.19: 42234664-42234664 on Build GRCh38
rs776647952 Chr.19: 42234574-42234574 on Build GRCh38
rs766460072 Chr.19: 42234627-42234627 on Build GRCh38
rs1020354740 Chr.19: 42234651-42234651 on Build GRCh38
rs761840963 Chr.19: 42234579-42234579 on Build GRCh38
rs749578178 Chr.19: 42234683-42234683 on Build GRCh38
rs535958519 Chr.19: 42234533-42234533 on Build GRCh38
rs865878103 Chr.19: 42234536-42234536 on Build GRCh38
rs746536605 Chr.19: 42234688-42234688 on Build GRCh38
rs539955255 Chr.19: 42234685-42234685 on Build GRCh38
rs773553059 Chr.19: 42234506-42234506 on Build GRCh38
rs199858279 Chr.19: 42234616-42234616 on Build GRCh38
rs554204519 Chr.19: 42234631-42234631 on Build GRCh38
rs754589092 Chr.19: 42234506-42234507 on Build GRCh38

View Full Product Details