Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 49952499-49952760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR4C13
Entrez Gene ID
283092
Gene Name
olfactory receptor family 4 subfamily C member 13
Gene Aliases
-
UniGene
Hs.553668
Gene Chromosome Location
Chr.11: 49952391-49953419 on Build GRCh38
Transcripts
NM_001001955.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1561376 c.251_268del18 p.Y85_I90delYENKTI Chr.11: 49952673-49952690 on Build GRCh38
4529537 c.161G>A p.R54K Chr.11: 49952583-49952583 on Build GRCh38
3448342 c.152C>T p.P51L Chr.11: 49952574-49952574 on Build GRCh38
4709893 c.106G>A p.A36T Chr.11: 49952528-49952528 on Build GRCh38
3448344 c.202G>A p.D68N Chr.11: 49952624-49952624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781939886 Chr.11: 49952587-49952587 on Build GRCh38
rs782349099 Chr.11: 49952579-49952579 on Build GRCh38
rs576355692 Chr.11: 49952649-49952649 on Build GRCh38
rs141959510 Chr.11: 49952548-49952548 on Build GRCh38
rs782181373 Chr.11: 49952527-49952527 on Build GRCh38
rs782628769 Chr.11: 49952704-49952704 on Build GRCh38
rs782593425 Chr.11: 49952713-49952713 on Build GRCh38
rs782692565 Chr.11: 49952554-49952554 on Build GRCh38
rs782720503 Chr.11: 49952544-49952544 on Build GRCh38
rs782469539 Chr.11: 49952696-49952696 on Build GRCh38
rs146392842 Chr.11: 49952726-49952726 on Build GRCh38
rs782759084 Chr.11: 49952596-49952596 on Build GRCh38
rs371893127 Chr.11: 49952550-49952550 on Build GRCh38
rs781828575 Chr.11: 49952672-49952672 on Build GRCh38
rs142197605 Chr.11: 49952716-49952716 on Build GRCh38
rs782275579 Chr.11: 49952559-49952559 on Build GRCh38
rs554023729 Chr.11: 49952592-49952592 on Build GRCh38
rs150235959 Chr.11: 49952659-49952659 on Build GRCh38
rs782258415 Chr.11: 49952642-49952642 on Build GRCh38
rs781856574 Chr.11: 49952619-49952619 on Build GRCh38
rs541814125 Chr.11: 49952650-49952650 on Build GRCh38
rs782486457 Chr.11: 49952625-49952625 on Build GRCh38
rs781954315 Chr.11: 49952597-49952597 on Build GRCh38
rs782398561 Chr.11: 49952588-49952588 on Build GRCh38
rs147550726 Chr.11: 49952545-49952545 on Build GRCh38
rs782530085 Chr.11: 49952567-49952567 on Build GRCh38
rs782442617 Chr.11: 49952632-49952632 on Build GRCh38
rs781835632 Chr.11: 49952708-49952708 on Build GRCh38
rs782462992 Chr.11: 49952709-49952709 on Build GRCh38
rs145904351 Chr.11: 49952553-49952553 on Build GRCh38
rs782372318 Chr.11: 49952645-49952645 on Build GRCh38
rs782632539 Chr.11: 49952568-49952568 on Build GRCh38
rs782383719 Chr.11: 49952535-49952535 on Build GRCh38
rs782064196 Chr.11: 49952603-49952603 on Build GRCh38
rs267602933 Chr.11: 49952703-49952703 on Build GRCh38
rs539709201 Chr.11: 49952628-49952628 on Build GRCh38
rs556799481 Chr.11: 49952646-49952646 on Build GRCh38
rs781854504 Chr.11: 49952549-49952549 on Build GRCh38
rs368329765 Chr.11: 49952542-49952542 on Build GRCh38
rs79794374 Chr.11: 49952617-49952617 on Build GRCh38
rs782663280 Chr.11: 49952634-49952634 on Build GRCh38
rs782268875 Chr.11: 49952734-49952734 on Build GRCh38
rs138947771 Chr.11: 49952698-49952698 on Build GRCh38
rs555352141 Chr.11: 49952684-49952684 on Build GRCh38
rs782706515 Chr.11: 49952697-49952697 on Build GRCh38
rs529445115 Chr.11: 49952574-49952574 on Build GRCh38
rs782767336 Chr.11: 49952666-49952666 on Build GRCh38
rs782202148 Chr.11: 49952631-49952631 on Build GRCh38
rs782376605 Chr.11: 49952735-49952735 on Build GRCh38
rs781907074 Chr.11: 49952614-49952614 on Build GRCh38
rs782113506 Chr.11: 49952652-49952652 on Build GRCh38
rs141161603 Chr.11: 49952533-49952533 on Build GRCh38
rs782431827 Chr.11: 49952731-49952731 on Build GRCh38
rs781994553 Chr.11: 49952591-49952591 on Build GRCh38
rs782090338 Chr.11: 49952543-49952543 on Build GRCh38
rs782259725 Chr.11: 49952596-49952596 on Build GRCh38
rs782799146 Chr.11: 49952651-49952651 on Build GRCh38
rs782031053 Chr.11: 49952641-49952641 on Build GRCh38
rs148216632 Chr.11: 49952605-49952605 on Build GRCh38
rs376714117 Chr.11: 49952528-49952528 on Build GRCh38

View Full Product Details