Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67433943-67434216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6KB2
Entrez Gene ID
6199
Gene Name
ribosomal protein S6 kinase B2
Gene Aliases
KLS, P70-beta, P70-beta-1, P70-beta-2, S6K-beta2, S6K2, SRK, STK14B, p70(S6K)-beta, p70S6Kb
UniGene
Hs.534345
Gene Chromosome Location
Chr.11: 67428460-67435408 on Build GRCh38
Transcripts
NM_003952.2 XM_005274164.1 XM_005274165.4 XM_006718655.3 XM_006718656.3 XM_006718657.1 XM_017018108.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6769600 c.985C>T p.R329W Chr.11: 67434213-67434213 on Build GRCh38
34768 c.986G>A p.R329Q Chr.11: 67434214-67434214 on Build GRCh38
6968485 c.938G>A p.G313E Chr.11: 67434026-67434026 on Build GRCh38
3869994 c.923C>T p.P308L Chr.11: 67434011-67434011 on Build GRCh38
6769596 c.962A>G p.D321G Chr.11: 67434050-67434050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759970102 Chr.11: 67434143-67434143 on Build GRCh38
rs750042369 Chr.11: 67434023-67434023 on Build GRCh38
rs752178018 Chr.11: 67434011-67434011 on Build GRCh38
rs996329859 Chr.11: 67434120-67434120 on Build GRCh38
rs747307396 Chr.11: 67434084-67434085 on Build GRCh38
rs201441323 Chr.11: 67434164-67434164 on Build GRCh38
rs749181013 Chr.11: 67433963-67433963 on Build GRCh38
rs569038614 Chr.11: 67434145-67434145 on Build GRCh38
rs779752969 Chr.11: 67434026-67434026 on Build GRCh38
rs771025035 Chr.11: 67434049-67434049 on Build GRCh38
rs2286623 Chr.11: 67433982-67433982 on Build GRCh38
rs753556578 Chr.11: 67434158-67434158 on Build GRCh38
rs1045320813 Chr.11: 67433975-67433975 on Build GRCh38
rs748658391 Chr.11: 67433976-67433977 on Build GRCh38
rs770985630 Chr.11: 67433973-67433973 on Build GRCh38
rs187977820 Chr.11: 67434024-67434024 on Build GRCh38
rs754738522 Chr.11: 67434188-67434188 on Build GRCh38
rs747101536 Chr.11: 67433969-67433969 on Build GRCh38
rs532185251 Chr.11: 67434100-67434100 on Build GRCh38
rs776029111 Chr.11: 67434004-67434004 on Build GRCh38
rs781156784 Chr.11: 67434067-67434067 on Build GRCh38
rs768582871 Chr.11: 67433965-67433965 on Build GRCh38
rs778732560 Chr.11: 67434190-67434190 on Build GRCh38
rs200667089 Chr.11: 67434019-67434019 on Build GRCh38
rs56326725 Chr.11: 67434144-67434144 on Build GRCh38
rs763397455 Chr.11: 67434096-67434096 on Build GRCh38
rs374309806 Chr.11: 67434155-67434155 on Build GRCh38
rs959667030 Chr.11: 67434121-67434121 on Build GRCh38
rs200357791 Chr.11: 67434150-67434150 on Build GRCh38
rs759728979 Chr.11: 67433990-67433990 on Build GRCh38
rs766984944 Chr.11: 67434020-67434020 on Build GRCh38
rs770236261 Chr.11: 67433991-67433991 on Build GRCh38
rs754822281 Chr.11: 67434029-67434029 on Build GRCh38
rs371599425 Chr.11: 67434138-67434138 on Build GRCh38
rs368101536 Chr.11: 67434076-67434076 on Build GRCh38
rs964150557 Chr.11: 67434108-67434108 on Build GRCh38
rs746064038 Chr.11: 67434073-67434073 on Build GRCh38
rs371472081 Chr.11: 67433982-67433983 on Build GRCh38
rs775941335 Chr.11: 67434081-67434081 on Build GRCh38
rs778820391 Chr.11: 67434030-67434030 on Build GRCh38
rs1029583414 Chr.11: 67434125-67434125 on Build GRCh38
rs201850616 Chr.11: 67434005-67434005 on Build GRCh38
rs777314327 Chr.11: 67433976-67433976 on Build GRCh38
rs777768823 Chr.11: 67433966-67433966 on Build GRCh38
rs773561013 Chr.11: 67434068-67434069 on Build GRCh38
rs753642062 Chr.11: 67434028-67434028 on Build GRCh38
rs748078979 Chr.11: 67434033-67434033 on Build GRCh38
rs769181434 Chr.11: 67434105-67434105 on Build GRCh38
rs772373231 Chr.11: 67433975-67433976 on Build GRCh38
rs774897455 Chr.11: 67434106-67434106 on Build GRCh38

View Full Product Details