Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1940687-1940960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSRB1
Entrez Gene ID
51734
Gene Name
methionine sulfoxide reductase B1
Gene Aliases
HSPC270, SELR, SELX, SEPX1, SepR
UniGene
Hs.655346
Gene Chromosome Location
Chr.16: 1938233-1943293 on Build GRCh38
Transcripts
NM_016332.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2129312 c.252C>T p.N84N Chr.16: 1940845-1940845 on Build GRCh38
5888288 c.265C>T p.P89S Chr.16: 1940832-1940832 on Build GRCh38
3672180 c.260C>A p.P87H Chr.16: 1940837-1940837 on Build GRCh38
6899476 c.267G>A p.P89P Chr.16: 1940830-1940830 on Build GRCh38
4058920 c.304A>C p.K102Q Chr.16: 1940793-1940793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771870935 Chr.16: 1940739-1940739 on Build GRCh38
rs764550663 Chr.16: 1940830-1940831 on Build GRCh38
rs774267686 Chr.16: 1940734-1940734 on Build GRCh38
rs552092161 Chr.16: 1940787-1940787 on Build GRCh38
rs368229562 Chr.16: 1940845-1940845 on Build GRCh38
rs141280253 Chr.16: 1940747-1940747 on Build GRCh38
rs772844550 Chr.16: 1940744-1940744 on Build GRCh38
rs755037727 Chr.16: 1940800-1940800 on Build GRCh38
rs763725376 Chr.16: 1940934-1940934 on Build GRCh38
rs752086291 Chr.16: 1940756-1940756 on Build GRCh38
rs770828953 Chr.16: 1940831-1940831 on Build GRCh38
rs755154134 Chr.16: 1940892-1940892 on Build GRCh38
rs971069567 Chr.16: 1940859-1940859 on Build GRCh38
rs750942303 Chr.16: 1940856-1940856 on Build GRCh38
rs950732653 Chr.16: 1940723-1940723 on Build GRCh38
rs753536485 Chr.16: 1940750-1940750 on Build GRCh38
rs376718791 Chr.16: 1940896-1940896 on Build GRCh38
rs757875054 Chr.16: 1940761-1940761 on Build GRCh38
rs746717797 Chr.16: 1940830-1940830 on Build GRCh38
rs920512270 Chr.16: 1940715-1940715 on Build GRCh38
rs775293204 Chr.16: 1940930-1940930 on Build GRCh38
rs779130498 Chr.16: 1940894-1940894 on Build GRCh38
rs371734249 Chr.16: 1940857-1940857 on Build GRCh38
rs748260206 Chr.16: 1940819-1940819 on Build GRCh38
rs781736264 Chr.16: 1940780-1940780 on Build GRCh38
rs762300391 Chr.16: 1940933-1940933 on Build GRCh38
rs759116211 Chr.16: 1940753-1940753 on Build GRCh38
rs534536139 Chr.16: 1940844-1940844 on Build GRCh38
rs765891991 Chr.16: 1940748-1940748 on Build GRCh38
rs769518673 Chr.16: 1940929-1940929 on Build GRCh38
rs761788187 Chr.16: 1940738-1940738 on Build GRCh38
rs779004449 Chr.16: 1940817-1940817 on Build GRCh38
rs766824835 Chr.16: 1940867-1940867 on Build GRCh38
rs773216710 Chr.16: 1940826-1940826 on Build GRCh38
rs764804277 Chr.16: 1940834-1940834 on Build GRCh38
rs540732571 Chr.16: 1940835-1940835 on Build GRCh38
rs775096631 Chr.16: 1940842-1940842 on Build GRCh38
rs759315403 Chr.16: 1940833-1940833 on Build GRCh38
rs771856577 Chr.16: 1940820-1940820 on Build GRCh38
rs368995810 Chr.16: 1940917-1940917 on Build GRCh38
rs754028633 Chr.16: 1940878-1940878 on Build GRCh38
rs749577355 Chr.16: 1940798-1940798 on Build GRCh38
rs776096819 Chr.16: 1940854-1940854 on Build GRCh38
rs750701559 Chr.16: 1940785-1940785 on Build GRCh38
rs758512081 Chr.16: 1940902-1940902 on Build GRCh38
rs374342988 Chr.16: 1940727-1940727 on Build GRCh38
rs761120034 Chr.16: 1940710-1940710 on Build GRCh38
rs780071439 Chr.16: 1940797-1940797 on Build GRCh38
rs777570742 Chr.16: 1940909-1940909 on Build GRCh38
rs773578369 Chr.16: 1940935-1940935 on Build GRCh38
rs565623090 Chr.16: 1940832-1940832 on Build GRCh38
rs770613695 Chr.16: 1940915-1940915 on Build GRCh38

View Full Product Details