Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 13977472-13977703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF182
Entrez Gene ID
221687
Gene Name
ring finger protein 182
Gene Aliases
-
UniGene
Hs.111164
Gene Chromosome Location
Chr.6: 13924446-13980009 on Build GRCh38
Transcripts
NM_001165032.1 NM_001165033.1 NM_001165034.1 NM_152737.3 XM_017010448.1 XM_017010449.1 XM_017010450.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1073914 c.451G>T p.E151* Chr.6: 13977570-13977570 on Build GRCh38
1073915 c.475G>A p.D159N Chr.6: 13977594-13977594 on Build GRCh38
3769718 c.543C>T p.S181S Chr.6: 13977662-13977662 on Build GRCh38
5624938 c.409C>A p.Q137K Chr.6: 13977528-13977528 on Build GRCh38
5658497 c.370A>G p.T124A Chr.6: 13977489-13977489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550428636 Chr.6: 13977577-13977577 on Build GRCh38
rs757107139 Chr.6: 13977521-13977521 on Build GRCh38
rs762477344 Chr.6: 13977677-13977677 on Build GRCh38
rs751355241 Chr.6: 13977514-13977514 on Build GRCh38
rs763771023 Chr.6: 13977593-13977593 on Build GRCh38
rs376459249 Chr.6: 13977666-13977666 on Build GRCh38
rs374399910 Chr.6: 13977509-13977509 on Build GRCh38
rs762522271 Chr.6: 13977589-13977589 on Build GRCh38
rs138868329 Chr.6: 13977559-13977559 on Build GRCh38
rs768950310 Chr.6: 13977637-13977637 on Build GRCh38
rs372439813 Chr.6: 13977500-13977500 on Build GRCh38
rs749382804 Chr.6: 13977621-13977621 on Build GRCh38
rs763717759 Chr.6: 13977679-13977679 on Build GRCh38
rs755389841 Chr.6: 13977525-13977525 on Build GRCh38
rs537106823 Chr.6: 13977567-13977567 on Build GRCh38
rs781214947 Chr.6: 13977527-13977527 on Build GRCh38
rs548450411 Chr.6: 13977552-13977552 on Build GRCh38
rs773576357 Chr.6: 13977659-13977659 on Build GRCh38
rs146832314 Chr.6: 13977640-13977640 on Build GRCh38
rs764858089 Chr.6: 13977581-13977581 on Build GRCh38
rs762290428 Chr.6: 13977560-13977560 on Build GRCh38
rs758172836 Chr.6: 13977496-13977496 on Build GRCh38
rs752212684 Chr.6: 13977586-13977586 on Build GRCh38
rs368857671 Chr.6: 13977667-13977667 on Build GRCh38
rs145658401 Chr.6: 13977643-13977643 on Build GRCh38
rs367962308 Chr.6: 13977608-13977608 on Build GRCh38
rs780225544 Chr.6: 13977619-13977619 on Build GRCh38
rs145031346 Chr.6: 13977594-13977594 on Build GRCh38
rs752410878 Chr.6: 13977491-13977491 on Build GRCh38
rs935163980 Chr.6: 13977495-13977496 on Build GRCh38
rs368233650 Chr.6: 13977499-13977499 on Build GRCh38
rs559717155 Chr.6: 13977544-13977544 on Build GRCh38
rs756136763 Chr.6: 13977615-13977615 on Build GRCh38
rs114884830 Chr.6: 13977641-13977641 on Build GRCh38
rs749580920 Chr.6: 13977547-13977547 on Build GRCh38
rs528284492 Chr.6: 13977545-13977545 on Build GRCh38
rs761183489 Chr.6: 13977665-13977665 on Build GRCh38
rs769013783 Chr.6: 13977558-13977558 on Build GRCh38
rs930018353 Chr.6: 13977507-13977507 on Build GRCh38
rs772434540 Chr.6: 13977561-13977561 on Build GRCh38

View Full Product Details