Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 16323297-16323534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXNAD1
Entrez Gene ID
92106
Gene Name
oxidoreductase NAD binding domain containing 1
Gene Aliases
-
UniGene
Hs.655449
Gene Chromosome Location
Chr.3: 16265160-16386979 on Build GRCh38
Transcripts
XM_011534232.2 XM_011534233.2 XM_011534234.2 XM_011534235.2 XM_017007485.1 XM_017007488.1 XM_017007489.1 XM_017007490.1 XM_017007491.1 XM_017007494.1 XM_017007495.1 XM_017007496.1 NM_015150.1 XM_005264985.1 XM_005264986.2 XM_006713069.1 XM_011533529.1 XM_011533530.1 XM_011533531.1 XM_017006005.1 XM_017006006.1 XM_017006007.1

Gene Symbol
RFTN1
Entrez Gene ID
23180
Gene Name
raftlin, lipid raft linker 1
Gene Aliases
MIG2, PIB10, PIG9, RAFTLIN
UniGene
Hs.98910
Gene Chromosome Location
Chr.3: 16315845-16513715 on Build GRCh38
Transcripts
XM_011534232.2 XM_011534233.2 XM_011534234.2 XM_011534235.2 XM_017007485.1 XM_017007488.1 XM_017007489.1 XM_017007490.1 XM_017007491.1 XM_017007494.1 XM_017007495.1 XM_017007496.1 NM_015150.1 XM_005264985.1 XM_005264986.2 XM_006713069.1 XM_011533529.1 XM_011533530.1 XM_011533531.1 XM_017006005.1 XM_017006006.1 XM_017006007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4463514 c.1295C>T p.P432L Chr.3: 16323413-16323413 on Build GRCh38
1040716 c.1329G>A p.S443S Chr.3: 16323379-16323379 on Build GRCh38
5736537 c.1323G>T p.K441N Chr.3: 16323385-16323385 on Build GRCh38
3589529 c.1256G>A p.G419E Chr.3: 16323452-16323452 on Build GRCh38
5398557 c.1290G>A p.Q430Q Chr.3: 16323418-16323418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994871775 Chr.3: 16323464-16323464 on Build GRCh38
rs1032638770 Chr.3: 16323448-16323448 on Build GRCh38
rs554929151 Chr.3: 16323469-16323469 on Build GRCh38
rs11554130 Chr.3: 16323447-16323447 on Build GRCh38
rs988838716 Chr.3: 16323372-16323372 on Build GRCh38
rs149141757 Chr.3: 16323473-16323473 on Build GRCh38
rs766277912 Chr.3: 16323435-16323435 on Build GRCh38
rs754232630 Chr.3: 16323456-16323456 on Build GRCh38
rs774327152 Chr.3: 16323439-16323439 on Build GRCh38
rs145153104 Chr.3: 16323360-16323360 on Build GRCh38
rs142197217 Chr.3: 16323457-16323457 on Build GRCh38
rs778211984 Chr.3: 16323389-16323389 on Build GRCh38
rs373734154 Chr.3: 16323367-16323367 on Build GRCh38
rs769430302 Chr.3: 16323429-16323429 on Build GRCh38
rs921377636 Chr.3: 16323490-16323490 on Build GRCh38
rs767056242 Chr.3: 16323449-16323449 on Build GRCh38
rs372038226 Chr.3: 16323484-16323484 on Build GRCh38
rs567669229 Chr.3: 16323342-16323342 on Build GRCh38
rs764763329 Chr.3: 16323385-16323387 on Build GRCh38
rs754328626 Chr.3: 16323475-16323477 on Build GRCh38
rs59223456 Chr.3: 16323339-16323339 on Build GRCh38
rs956517622 Chr.3: 16323513-16323513 on Build GRCh38
rs773026985 Chr.3: 16323430-16323430 on Build GRCh38
rs755812571 Chr.3: 16323453-16323453 on Build GRCh38
rs779289711 Chr.3: 16323420-16323420 on Build GRCh38
rs780208556 Chr.3: 16323470-16323470 on Build GRCh38
rs775509662 Chr.3: 16323415-16323415 on Build GRCh38
rs200833904 Chr.3: 16323455-16323455 on Build GRCh38
rs765839281 Chr.3: 16323363-16323363 on Build GRCh38
rs775095631 Chr.3: 16323509-16323509 on Build GRCh38
rs180950245 Chr.3: 16323391-16323391 on Build GRCh38
rs1057490805 Chr.3: 16323413-16323413 on Build GRCh38
rs149272972 Chr.3: 16323379-16323379 on Build GRCh38
rs762904260 Chr.3: 16323358-16323358 on Build GRCh38
rs756365890 Chr.3: 16323387-16323387 on Build GRCh38
rs947354084 Chr.3: 16323380-16323380 on Build GRCh38
rs953241634 Chr.3: 16323331-16323331 on Build GRCh38
rs770776552 Chr.3: 16323477-16323477 on Build GRCh38
rs372312509 Chr.3: 16323468-16323468 on Build GRCh38
rs747163191 Chr.3: 16323471-16323471 on Build GRCh38
rs1028655937 Chr.3: 16323465-16323465 on Build GRCh38
rs762772839 Chr.3: 16323431-16323431 on Build GRCh38
rs375698035 Chr.3: 16323475-16323475 on Build GRCh38
rs751034312 Chr.3: 16323365-16323365 on Build GRCh38
rs368712393 Chr.3: 16323406-16323406 on Build GRCh38
rs759528620 Chr.3: 16323487-16323487 on Build GRCh38
rs771595914 Chr.3: 16323404-16323404 on Build GRCh38
rs690216 Chr.3: 16323412-16323412 on Build GRCh38
rs760967852 Chr.3: 16323416-16323416 on Build GRCh38

View Full Product Details