Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218073206-218073431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RUFY4
Entrez Gene ID
285180
Gene Name
RUN and FYVE domain containing 4
Gene Aliases
ZFYVE31
UniGene
Hs.570069
Gene Chromosome Location
Chr.2: 218034934-218090584 on Build GRCh38
Transcripts
NM_198483.3 NR_034176.1 XM_011511014.2 XM_017003895.1 XM_017003896.1 XM_017003897.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1016278 c.512C>T p.A171V Chr.2: 218073368-218073368 on Build GRCh38
4803123 c.427C>T p.R143C Chr.2: 218073283-218073283 on Build GRCh38
5445248 c.460C>A p.L154I Chr.2: 218073316-218073316 on Build GRCh38
5538430 c.388G>A p.E130K Chr.2: 218073244-218073244 on Build GRCh38
1016279 c.512C>T p.A171V Chr.2: 218073368-218073368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372544844 Chr.2: 218073390-218073390 on Build GRCh38
rs913005670 Chr.2: 218073367-218073367 on Build GRCh38
rs1033197790 Chr.2: 218073343-218073343 on Build GRCh38
rs897236777 Chr.2: 218073277-218073277 on Build GRCh38
rs749444910 Chr.2: 218073320-218073320 on Build GRCh38
rs764471842 Chr.2: 218073394-218073394 on Build GRCh38
rs771870870 Chr.2: 218073334-218073334 on Build GRCh38
rs773012062 Chr.2: 218073303-218073306 on Build GRCh38
rs55862020 Chr.2: 218073333-218073333 on Build GRCh38
rs1047356067 Chr.2: 218073226-218073226 on Build GRCh38
rs931917697 Chr.2: 218073258-218073258 on Build GRCh38
rs888461089 Chr.2: 218073264-218073264 on Build GRCh38
rs764924187 Chr.2: 218073377-218073377 on Build GRCh38
rs1037140552 Chr.2: 218073269-218073269 on Build GRCh38
rs781519802 Chr.2: 218073287-218073287 on Build GRCh38
rs765974891 Chr.2: 218073244-218073244 on Build GRCh38
rs757731568 Chr.2: 218073286-218073286 on Build GRCh38
rs758983979 Chr.2: 218073254-218073254 on Build GRCh38
rs766239038 Chr.2: 218073383-218073384 on Build GRCh38
rs201339003 Chr.2: 218073361-218073361 on Build GRCh38
rs757642023 Chr.2: 218073402-218073402 on Build GRCh38
rs200798920 Chr.2: 218073349-218073349 on Build GRCh38
rs752316048 Chr.2: 218073260-218073260 on Build GRCh38
rs746101932 Chr.2: 218073292-218073292 on Build GRCh38
rs1040197263 Chr.2: 218073249-218073249 on Build GRCh38
rs867379955 Chr.2: 218073374-218073374 on Build GRCh38
rs752134165 Chr.2: 218073400-218073400 on Build GRCh38
rs776288032 Chr.2: 218073246-218073246 on Build GRCh38
rs747485182 Chr.2: 218073357-218073357 on Build GRCh38
rs1005542026 Chr.2: 218073267-218073267 on Build GRCh38
rs900032097 Chr.2: 218073304-218073304 on Build GRCh38
rs774838402 Chr.2: 218073385-218073385 on Build GRCh38
rs184111005 Chr.2: 218073311-218073311 on Build GRCh38
rs528005712 Chr.2: 218073318-218073318 on Build GRCh38
rs551024765 Chr.2: 218073276-218073276 on Build GRCh38
rs762752408 Chr.2: 218073310-218073311 on Build GRCh38
rs542772060 Chr.2: 218073259-218073259 on Build GRCh38
rs565548343 Chr.2: 218073252-218073252 on Build GRCh38
rs758815651 Chr.2: 218073278-218073278 on Build GRCh38
rs759180219 Chr.2: 218073372-218073372 on Build GRCh38
rs773231038 Chr.2: 218073335-218073335 on Build GRCh38
rs199659199 Chr.2: 218073284-218073284 on Build GRCh38
rs957280710 Chr.2: 218073407-218073407 on Build GRCh38
rs569550024 Chr.2: 218073283-218073283 on Build GRCh38
rs767998455 Chr.2: 218073404-218073404 on Build GRCh38

View Full Product Details