Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 84888611-84888860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC28A1
Entrez Gene ID
9154
Gene Name
solute carrier family 28 member 1
Gene Aliases
CNT1, HCNT1
UniGene
Hs.459187
Gene Chromosome Location
Chr.15: 84884351-84975649 on Build GRCh38
Transcripts
NM_001287761.1 NM_001287762.1 NM_001321721.1 NM_001321722.1 NM_004213.4 NM_201651.2 XM_011522203.2 XM_011522204.2 XM_011522205.2 XM_011522206.2 XM_011522207.2 XM_011522208.2 XM_011522209.2 XM_011522210.2 XM_011522211.2 XM_011522212.1 XM_011522214.2 XM_011522215.2 XM_011522216.2 XM_011522217.1 XM_011522218.2 XM_017022723.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4459554 c.112C>T p.P38S Chr.15: 84888787-84888787 on Build GRCh38
5588956 c.138G>T p.E46D Chr.15: 84888813-84888813 on Build GRCh38
1629720 c.162G>A p.A54A Chr.15: 84888837-84888837 on Build GRCh38
5588957 c.138G>T p.E46D Chr.15: 84888813-84888813 on Build GRCh38
6407858 c.115A>G p.R39G Chr.15: 84888790-84888790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781301587 Chr.15: 84888837-84888837 on Build GRCh38
rs960723800 Chr.15: 84888669-84888669 on Build GRCh38
rs753472790 Chr.15: 84888800-84888800 on Build GRCh38
rs778713664 Chr.15: 84888763-84888763 on Build GRCh38
rs990603225 Chr.15: 84888684-84888684 on Build GRCh38
rs17215892 Chr.15: 84888807-84888807 on Build GRCh38
rs760317644 Chr.15: 84888796-84888796 on Build GRCh38
rs543809231 Chr.15: 84888752-84888752 on Build GRCh38
rs375692167 Chr.15: 84888832-84888832 on Build GRCh38
rs1010831369 Chr.15: 84888757-84888757 on Build GRCh38
rs17222302 Chr.15: 84888799-84888799 on Build GRCh38
rs999808003 Chr.15: 84888776-84888776 on Build GRCh38
rs778321548 Chr.15: 84888831-84888831 on Build GRCh38
rs778192647 Chr.15: 84888744-84888744 on Build GRCh38
rs538274620 Chr.15: 84888816-84888816 on Build GRCh38
rs779300729 Chr.15: 84888721-84888721 on Build GRCh38
rs376752363 Chr.15: 84888680-84888680 on Build GRCh38
rs541757744 Chr.15: 84888726-84888726 on Build GRCh38
rs768377268 Chr.15: 84888772-84888772 on Build GRCh38
rs17215920 Chr.15: 84888703-84888703 on Build GRCh38
rs750912996 Chr.15: 84888692-84888692 on Build GRCh38
rs767578621 Chr.15: 84888788-84888788 on Build GRCh38
rs761652882 Chr.15: 84888787-84888787 on Build GRCh38
rs17222295 Chr.15: 84888803-84888803 on Build GRCh38
rs1040597972 Chr.15: 84888740-84888740 on Build GRCh38
rs1003427484 Chr.15: 84888670-84888670 on Build GRCh38
rs113577701 Chr.15: 84888838-84888838 on Build GRCh38
rs907378857 Chr.15: 84888653-84888653 on Build GRCh38
rs1045005959 Chr.15: 84888652-84888652 on Build GRCh38
rs1031405055 Chr.15: 84888693-84888693 on Build GRCh38
rs35565153 Chr.15: 84888696-84888697 on Build GRCh38
rs371089626 Chr.15: 84888784-84888784 on Build GRCh38
rs17215899 Chr.15: 84888729-84888729 on Build GRCh38
rs1011228909 Chr.15: 84888702-84888702 on Build GRCh38
rs17222316 Chr.15: 84888750-84888750 on Build GRCh38
rs978639655 Chr.15: 84888656-84888656 on Build GRCh38
rs375042078 Chr.15: 84888840-84888840 on Build GRCh38
rs985402023 Chr.15: 84888736-84888736 on Build GRCh38
rs8187742 Chr.15: 84888762-84888762 on Build GRCh38
rs930255137 Chr.15: 84888701-84888701 on Build GRCh38
rs777233521 Chr.15: 84888839-84888839 on Build GRCh38
rs572528168 Chr.15: 84888725-84888725 on Build GRCh38
rs180875143 Chr.15: 84888792-84888792 on Build GRCh38
rs201974180 Chr.15: 84888836-84888836 on Build GRCh38
rs940642665 Chr.15: 84888738-84888738 on Build GRCh38
rs928176357 Chr.15: 84888665-84888665 on Build GRCh38
rs538862956 Chr.15: 84888641-84888641 on Build GRCh38
rs984829867 Chr.15: 84888790-84888790 on Build GRCh38
rs1013167485 Chr.15: 84888636-84888636 on Build GRCh38
rs141245994 Chr.15: 84888805-84888805 on Build GRCh38
rs536036341 Chr.15: 84888822-84888822 on Build GRCh38
rs754536603 Chr.15: 84888753-84888753 on Build GRCh38

View Full Product Details