Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57231542-57231787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHMT2
Entrez Gene ID
6472
Gene Name
serine hydroxymethyltransferase 2
Gene Aliases
GLYA, HEL-S-51e, SHMT
UniGene
Hs.741179
Gene Chromosome Location
Chr.12: 57229573-57234935 on Build GRCh38
Transcripts
NM_001166356.1 NM_001166357.1 NM_001166358.1 NM_001166359.1 NM_005412.5 NR_029415.1 NR_029416.1 NR_029417.1 NR_048562.1 XM_011538675.2 XM_011538676.2 XM_011538677.2 XM_011538678.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7299363 c.364C>T p.R122W Chr.12: 57231765-57231765 on Build GRCh38
4834658 c.12G>C p.Q4H Chr.12: 57231729-57231729 on Build GRCh38
941997 c.367G>A p.A123T Chr.12: 57231768-57231768 on Build GRCh38
6777901 c.45C>T p.S15S Chr.12: 57231762-57231762 on Build GRCh38
1988705 c.312-6C>T p.? Chr.12: 57231707-57231707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376129607 Chr.12: 57231584-57231584 on Build GRCh38
rs781143002 Chr.12: 57231720-57231720 on Build GRCh38
rs746120843 Chr.12: 57231756-57231756 on Build GRCh38
rs868128613 Chr.12: 57231749-57231749 on Build GRCh38
rs543579719 Chr.12: 57231767-57231767 on Build GRCh38
rs768352054 Chr.12: 57231634-57231634 on Build GRCh38
rs200146345 Chr.12: 57231607-57231607 on Build GRCh38
rs749441303 Chr.12: 57231598-57231598 on Build GRCh38
rs781045017 Chr.12: 57231592-57231592 on Build GRCh38
rs150389862 Chr.12: 57231714-57231714 on Build GRCh38
rs758782130 Chr.12: 57231757-57231757 on Build GRCh38
rs375584473 Chr.12: 57231762-57231762 on Build GRCh38
rs769162322 Chr.12: 57231677-57231677 on Build GRCh38
rs762502537 Chr.12: 57231688-57231688 on Build GRCh38
rs921546995 Chr.12: 57231633-57231633 on Build GRCh38
rs370553351 Chr.12: 57231612-57231612 on Build GRCh38
rs745814583 Chr.12: 57231752-57231752 on Build GRCh38
rs747439440 Chr.12: 57231763-57231763 on Build GRCh38
rs774000145 Chr.12: 57231705-57231705 on Build GRCh38
rs754006426 Chr.12: 57231734-57231734 on Build GRCh38
rs758393060 Chr.12: 57231666-57231667 on Build GRCh38
rs764074961 Chr.12: 57231722-57231722 on Build GRCh38
rs765960509 Chr.12: 57231699-57231699 on Build GRCh38
rs779210015 Chr.12: 57231753-57231753 on Build GRCh38
rs776096635 Chr.12: 57231672-57231672 on Build GRCh38
rs747569156 Chr.12: 57231675-57231675 on Build GRCh38
rs968967684 Chr.12: 57231648-57231648 on Build GRCh38
rs756098247 Chr.12: 57231595-57231595 on Build GRCh38
rs771249894 Chr.12: 57231606-57231606 on Build GRCh38
rs752663968 Chr.12: 57231593-57231593 on Build GRCh38
rs373976238 Chr.12: 57231686-57231686 on Build GRCh38
rs373757480 Chr.12: 57231707-57231707 on Build GRCh38
rs767322828 Chr.12: 57231708-57231708 on Build GRCh38
rs11557162 Chr.12: 57231738-57231738 on Build GRCh38
rs1040468983 Chr.12: 57231623-57231623 on Build GRCh38
rs777960534 Chr.12: 57231597-57231597 on Build GRCh38
rs978885937 Chr.12: 57231655-57231655 on Build GRCh38
rs944806740 Chr.12: 57231620-57231620 on Build GRCh38

View Full Product Details